Skip to Content
Merck
All Photos(1)

Key Documents

MAB8906

Sigma-Aldrich

Anti-Measles Antibody, nucleoprotein, clone 83KKII

clone 83KKII, Chemicon®, from mouse

Synonym(s):

Rubeola

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

83KKII, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunofluorescence: suitable

isotype

IgG

shipped in

wet ice

General description

Measles is a highly contagious, acute febrile illness that has been one of the most common viral diseases of young children. Although measles has been effectively controlled in recent years by the use of live attenuated virus vaccines, measles continues to have a serious impact on human health as evidenced by its continued persistence in developing countries. Measles is characterized by a prodrome illness with rapidly rising fever that culminates in a generalized macular-papular rash. The appearance of Koplik′s spots on the mouth epithelium during or after the prodromal phase is pathognomonic for measles. The measles virus belongs to the family Paramyxoviridae, and is a member of the genus Morbillivirus. Measles is an enveloped, pleomorphic virus ranging from 100 to 250 nm in diameter with a single stranded linear RNA. Infected cells contain large amounts of nucleoprotein antigen which is the major internal protein of measles virus. Consequently, the dominant part of the antibody response to measles virus involves the nucleoprotein antigen.

Specificity

MAB8906 recognizes the measles nucleoprotein by capture EIA.

Immunogen

Epitope: Nucleoprotein

Application

Capture ELISA: A 1:5,000-1:10,000 dilution of a previous lot was used in ELISA.

Indirect Immunofluorescence: A previous lot of this antibody was used on indirect immunofluorescence.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected the measles nucleoprotein in cells infected with measles virus.

Optimal working dilutions must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral
This Anti-Measles Antibody, nucleoprotein, clone 83KKII is validated for use in ELISA, IF for the detection of Measles.

Physical form

Format: Purified
Protein A purified
Purified mouse monoclonal IgG in buffer containing liquid in PBS with 0.1% sodium azide.

Storage and Stability

Stable for 1 year at from date of receipt.

Analysis Note

Control
Measles Control Slides, Catalogue Number 5031-5

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lung involvement in childhood measles: severe immune dysfunction revealed by quantitative immunohistochemistry.
Talib Moyses Moussallem,Fernanda Guedes,Elaine Raniero Fernandes,Carla Pagliari et al.
Human Pathology null
Measles virus entry through the signaling lymphocyte activation molecule governs efficacy of mantle cell lymphoma radiovirotherapy.
Miest, TS; Frenzke, M; Cattaneo, R
Molecular Therapy null
Adherens junction protein nectin-4 is the epithelial receptor for measles virus.
Muhlebach, MD; Mateo, M; Sinn, PL; Prufer, S; Uhlig, KM; Leonard, VH; Navaratnarajah et al.
Nature null
Daria Jacob et al.
PloS one, 9(1), e86658-e86658 (2014-01-30)
Yeasts are largely used as bioreactors for vaccine production. Usually, antigens are produced in yeast then purified and mixed with adjuvants before immunization. However, the purification costs and the safety concerns recently raised by the use of new adjuvants argue
Immunohistochemical detection of virus through its nuclear cytopathic effect in idiopathic interstitial pneumonia other than acute exacerbation.
Santos, GC; Parra, ER; Stegun, FW; Cirqueira, CS; Capelozzi, VL
Brazilian journal of medical and biological research null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service