Skip to Content
Merck
All Photos(1)

Key Documents

MAB8535

Sigma-Aldrich

Anti-Herpes Virus 6 Antibody, 101kDa protein, clone C3108-103

clone C3108-103, Chemicon®, from mouse

Synonym(s):

HHV-6B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

C3108-103, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

shipped in

wet ice

Specificity

Human herpesvirus 6, B variant. Recognizes a viron protein of 101 kDa.

SPECIES REACTIVITIES:

Shows weak reactivity to cells infected with human CMV in immunofluorescent assays. Does not cross-react with HHV-6 A variant.

Immunogen

Epitope: 101kDa protein

Application

Indirect immunofluorescence at 1:50+ (infected culture cells).

Immunohistochemistry, IH/IH(P)

Dilute with buffer pH 7.5-8.0 to desired working volume.

For extensive dilution, protein containing or other stabilizing medium should be used.

Final working dilutions must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral
This Anti-Herpes Virus 6 Antibody, 101kDa protein, clone C3108-103 is validated for use in IF, WB, IH for the detection of Herpes Virus 6.

Physical form

Format: Purified
In 0.02M phoshate buffer, containing 0.25M NaCl and 0.1% Sodium Azide, pH 7.6.

Storage and Stability

Maintain at 2-8°C for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human herpesvirus 6 and cytomegalovirus in ileocolonic mucosa in inflammatory bowel disease.
Taina Sipponen,Ulla Turunen,Irmeli Lautenschlager,Urpo Nieminen,Johanna Arola,Leena Halme
Scandinavian Journal of Gastroenterology null
HHV-6 and HHV-7 antigenemia related to CMV infection after liver transplantation.
Maiju Harma, Krister Hockerstedt, Outi Lyytikainen, Irmeli Lautenschlager
Journal of Medical Virology null
Human herpesvirus 6 antigenaemia in allogeneic stem cell transplant recipients: impact on clinical course and association with other beta-herpesviruses.
Liisa Volin, Irmeli Lautenschlager, Eeva Juvonen, Anne Nihtinen, Veli-Jukka Anttila, Tapani Ruutu
British Journal of Haematology null
Quantitative HHV-6B antigenemia test for the monitoring of transplant patients.
R Loginov,T Karlsson,K Hockerstedt,D Ablashi,I Lautenschlager
European Journal of Clinical Microbiology & Infectious Diseases : Official Publication of the European Society of Clinical Microbiology null
Ana Lia Pradella Puglia et al.
Revista da Sociedade Brasileira de Medicina Tropical, 53, e20190181-e20190181 (2020-03-19)
Human herpesvirus (HHV)-7 establishes a latent infection during the lifetime of the host and can reactivate after the primary infection, leading to lytic replication in immunosuppressed patients. This study aimed to develop an enzyme-linked immunosorbent assay (ELISA) to identify HHV-7

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service