Skip to Content
Merck
All Photos(8)

Key Documents

MAB5284

Sigma-Aldrich

Anti-Chondroitin Sulfate Proteoglycan Antibody, Brain (core protein), clone Cat-301

clone Cat-301, Chemicon®, from mouse

Synonym(s):

CSPG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

Cat-301, monoclonal

species reactivity

primate, human, rat, gerbil, feline, hamster, guinea pig

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ACAN(176)

General description

The antigen recognized by this clone, CAT-301 is a proteoglycan that is developmentaly regulated, high molecular chondroitin sulphate proteoglycan found on the extracellular surface of mammalian neurons. It is expressed late in development and although no definitive role has been identified for CAT-301, it is believed to have a role in the stabilisation of synaptic structure.

The name derives from the name of the monoclonal antibody originally used to identified by its expression, which was detected immunocytochemicaly



Disruption of the normal patterns of neuronal activity during the critical early postnatal period by physical or biochemical means results in a large and irreversible reduction in levels of CAT-301. Similar intervention in mature animals has no effect



CAT-301 is related to aggrecan from articular cartilage. The CAT-301 antibody immunoreacts with aggrecan, and the chondroitinase treated aggrecan and CAT-301 both have apparent sizes of 550kDa. However CAT- 301 is not glycosylated by keratan sulphate, as aggrecan is.

Specificity

Chondroitin sulfate proteoglycan (CSPG), brain core protein.

Immunogen

Epitope: Brain (core protein)
Feline spinal cord fixed gray matter.

Application

Anti-Chondroitin Sulfate Proteoglycan Antibody, Brain (core protein), clone Cat-301 detects level of Chondroitin Sulfate Proteoglycan & has been published & validated for use in IP, WB, IC, IH.
Immunohistochemistry: 1:500-1:2,000 on 4% paraformaldehyde fixed frozen tissue.

Immunocytochemistry

Immunoblot

Immunoprecipitation

Optimal working dilutions must be determined by the end user.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lena Iwai et al.
Cerebral cortex (New York, N.Y. : 1991), 23(9), 2204-2212 (2012-07-14)
Although the parallel visual pathways are a fundamental basis of visual processing, our knowledge of their molecular properties is still limited. Here, we uncovered a parvocellular-specific molecule in the dorsal lateral geniculate nucleus (dLGN) of higher mammals. We found that
Otavio S C Mariani et al.
The Journal of comparative neurology, 527(3), 694-717 (2018-03-27)
We propose a partitioning of the primate intraparietal sulcus (IPS) using immunoarchitectural and connectivity criteria. We studied the immunoarchitecture of the IPS areas in the capuchin monkey using Cat-301 and SMI-32 immunohistochemistry. In addition, we investigated the IPS projections to
Maija-Kreetta Koskinen et al.
Scientific reports, 10(1), 17308-17308 (2020-10-16)
Stress can predispose to depressive episodes, yet the molecular mechanisms regulating the transition from the initial stress response to a persistent pathological depressive state remain poorly understood. We profiled the development of an enduring depressive-like state by assessing affective behavior
Vasily Vorobyov et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(1), 234-243 (2013-01-04)
Monocular deprivation (MD) during a critical period of postnatal development produces significant changes in the anatomy and physiology of the visual cortex, and the deprived eye becomes amblyopic. Extracellular matrix molecules have a major role in restricting plasticity such that
Daniel L Felch et al.
Frontiers in neuroanatomy, 6, 12-12 (2012-04-20)
Previous research has suggested that the three physiologically defined relay cell-types in mammalian lateral geniculate nucleus (LGN)-called parvocellular (P), magnocellular (M), and koniocellular (K) cells in primates and X, Y, and W cells in other mammals-each express a unique combination

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service