Skip to Content
Merck
All Photos(1)

Key Documents

MAB4163

Sigma-Aldrich

Anti-MDR1 Antibody, extracellular human specific Pgp, clone MC57

clone MC57, Chemicon®, from mouse

Synonym(s):

P-glycoprotein, CD243, p170, Pgp

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.42

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MC57, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... ABCB1(5243)

Related Categories

Specificity

MDR-1 clone MC57 recgonizes the inactivated form of Pgp and recognizes the conformational epitope in Immunohistochemistry on paraffin sections.

Immunogen

Epitope: extracellular human specific Pgp
Living intact human multidrug-resistant cells

Application

Anti-MDR1 Antibody, extracellular human specific Pgp, clone MC57 detects level of MDR1 & has been published & validated for use in FC, IH, IH(P).
Flow Cytometry

Immunohistochemistry

Immunohistochemistry on paraffin sections

Functional Experiments

Optimal working dilutions must be determined by end user.
Research Category
Metabolism
Research Sub Category
Toxicology & Drug Resistance

Physical form

Format: Purified
Purified immunoglobulin from Protein A Sepharose chromatography. Liquid in 0.02M PB, pH 7.6, 0.25M NaCl containing 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 6 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsai-Yu Lee et al.
Oncotarget, 7(17), 23512-23520 (2016-03-19)
A tobacco-specific component, 4-methylnitrosamino-1-3-pyridyl-1-butanone (NNK), is a major risk factor for many cancers. Recent reports have demonstrated that NNK exposure may be associated with tumor progression and chemoresistance in certain cancers. However, the underlying NNK-induced mechanism contributing to the aggressiveness

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service