Skip to Content
Merck
All Photos(1)

Key Documents

MAB3510

Sigma-Aldrich

Anti-BrdU Antibody, clone BU-1

culture supernatant, clone BU-1, Chemicon®

Synonym(s):

BrdU

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

BU-1, monoclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable

isotype

IgG2a

shipped in

wet ice

target post-translational modification

unmodified

General description

Bromodeoxyuridine (BrdU) is a thymidine analog that, when added exogenously, is incorporated into DNA during DNA synthesis. Anti-bromodeoxyuridine monoclonal antibody is used to label proliferating cells that have incorporated BrdU. This immunological detection scheme has several advantages over the use of radioactive thymidine incorporation for identifying cells undergoing replication. Labeling and detection can be performed the same day instead of waiting several days, as required for autoradiography of tritium-labeled cells, and the necessity of using multiple specimens for obtaining the optimal exposure time is eliminated. In addition, anti-bromodeoxyuridine staining with flow cytometric analysis allows multiple parameters to be evaluated simultaneously.

Specificity

Clone BU-1 specifically binds to bromodeoxyuridine with 10% crossreactivity toward iodouridine (10%). Clone BU-1 does not crossreact with fluorodeoxyuridine or any endogenous cellular components such as thymidine or uridine.

Immunogen

Bromodeoxyuridine
Epitope: BrdU

Application

Although originally reported to stain nuclear DNA BrdU incorporation in ethanol-fixed cells without acid or base denaturation, this property was later attributed to the DNase activity present in a mycoplasma-contaminated BU-1 hybridoma culture supernatant used (Gonchoroff, N.J., et al. (1986). J. Immunol. Methods. 93(1):97-101; Greipp, P.R., et al. (1985). Am. J. Hematol. 20(3):289-292). Please include an appropriate DNA denaturation step during sample preparation prior to immunostaining using this product.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair
Use mouse monoclonal Anti-BrdU Antibody, clone BU-1, validated in Flow Cytometry, Immunocytochemistry, and Immunohistochemistry for the detection of bromodeoxyuridine (BrdU).

Physical form

Hybridoma culture supernatant from a perfusion system, filtered through a 0.2 micron membrane prior to vialing. Product contains 20% FBS and Ciprofloxacin at a final concentration of 10 μg/mL. Possible mycoplasma risk contained within this product. Use appropriate measures to prevent cell line infection/contamination.
Unpurified

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Brdu treated cells

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hypoxia-ischemia induces an endogenous reparative response by local neural progenitors in the postnatal mouse telencephalon.
Dizon, M; Szele, F; Kessler, JA
Developmental Neuroscience null
Miwako Kobayashi et al.
Molecular brain, 7, 12-12 (2014-02-18)
We have previously identified BRINP (BMP/RA-inducible neural-specific protein-1, 2, 3) family genes that possess the ability to suppress cell cycle progression in neural stem cells. Of the three family members, BRINP1 is the most highly expressed in various brain regions
Jessica Dragotto et al.
Journal of applied genetics, 60(3-4), 357-365 (2019-09-06)
The Npc1nih/nih-null model and the Npc1nmf164/nmf164 hypomorph models of Niemann-Pick C1 (NPC1) disease show defects in olfaction. We have tested the effects of the life-prolonging treatment hydroxypropyl-beta-cyclodextrin (HPBCD) on olfaction and neural stem cell numbers when delivered either systemically or
Leonid A Serebryannyy et al.
Journal of cell science, 129(18), 3412-3425 (2016-08-10)
Actin is abundant in the nucleus and it is clear that nuclear actin has important functions. However, mystery surrounds the absence of classical actin filaments in the nucleus. To address this question, we investigated how polymerizing nuclear actin into persistent
Differential involvement of estrogen receptor alpha and estrogen receptor beta in the healing promoting effect of estrogen in human keratinocytes.
Merlo, S; Frasca, G; Canonico, PL; Sortino, MA
The Journal of Endocrinology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service