Skip to Content
Merck
All Photos(1)

Key Documents

MAB3446

Sigma-Aldrich

Anti-Heterochromatin Protein-1 α Antibody, clone 2HP-2G9

ascites fluid, clone 2HP-2G9, Chemicon®

Synonym(s):

HP1a, Antigen p25, Chromobox Protein Homolog 5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

2HP-2G9, monoclonal

species reactivity

mouse, human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CBX5(23468)

General description

Chromatin assembly factor-1 (CAF-1) is a multisubunit protein complex that comprises three polypeptide subunits known as p150, p60, and p48. CAF-1 is a nucleosome assembly factor that deposits newly synthesized and acetylated histones H3/H4 into nascent chromatin during DNA replication.

Heterochromatin is characterized as densely coiled chromatin that generally replicates late during S phase, has a low gene density, and contains large blocks of repetitive DNA that is relatively inaccessible to DNA-modifying reagents. In late S phase, p150 directly associates with heterochromatin associated proteins 1 (HP1α, HP1β and HP1γ). As cells prepare for mitosis, CAF-1 p150 and some HP1 progressively dissociate from heterochromatin, coinciding with the phosphorylation of histone H3. The HP1 proteins reassociate with chromatin at the end of mitosis, as histone H3 is dephosphorylated.

Specificity

Reacts with Heterochromatin protein-1 alpha (HP1alpha) between amino acids 67 and 119.

Immunogen

Recombinant mouse HP1alpha.

Application

Anti-Heterochromatin Protein-1 α Antibody, clone 2HP-2G9 is a Mouse Monoclonal Antibody for detection of Heterochromatin Protein-1 α also known as HP1a, Antigen p25, Chromobox Protein Homolog 5 & has been validated in ELISA, IP & WB.
ELISA: 1:500-1:5,000

Western blot: 1:500-1:5,000

Immunoprecipitation: 1:500-1:5,000

Immunohistochemistry/Immunocytochemistry: Not Recommended

Note: for immunocytochemistry and immunohistochemistry we recommend CHEMICON catalog number MAB3584.

Optimal working dilutions must be determined by the end user.

Target description

25 kDa

Physical form

Liquid ascites fluid containing no preservative.

Analysis Note

Control
Positive Control: HeLa cells, 3T3 cells , F9 cells, mouse embryos

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A role for heterochromatin protein 1? at human telomeres.
Canudas, S; Houghtaling, BR; Bhanot, M; Sasa, G; Savage, SA; Bertuch, AA; Smith, S
Genes & Development null
Aurora-B/AIM-1 regulates the dynamic behavior of HP1alpha at the G2-M transition.
Terada, Y
Molecular Biology of the Cell null
Xiaoyuan Wei et al.
Journal of molecular cell biology, 6(5), 409-420 (2014-05-20)
The revolutionary induced pluripotent stem cell (iPSC) technology provides a new means for cell replacement therapies and drug screening. Small molecule compounds have been found extremely useful to improve the generation of iPSCs and understand the reprogramming mechanism. Here we
Distinctive patterns of epigenetic marks are associated with promoter regions of mouse LINE-1 and LTR retrotransposons.
Rangasamy, D
Mobile DNA null
Andrey Poleshko et al.
Journal of virology, 82(5), 2313-2323 (2007-12-21)
Integrated retroviral DNA is subject to epigenetic gene silencing, resulting in loss of expression of viral genes as well as reporter or therapeutic genes transduced by retroviral vectors. Possible mediators of such silencing include the histone deacetylase (HDAC) family of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service