Skip to Content
Merck
All Photos(1)

Key Documents

MAB3232

Sigma-Aldrich

Anti-Cytokeratin 14 Antibody, clone RCK107

clone RCK107, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RCK107, monoclonal

species reactivity

rat, pig, human, canine

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG3

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... KRT14(3861)
rat ... Krt14(287701)

General description

Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in epithelial tissues by at least 20 different polypeptides. They range in molecular weight from between 40 kDa and 68 kDa and isoelectric pH between 4.9 - 7.8, The individual cytokeratin polypeptides are designated 1-20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies.

Specificity

Cytokeratin 14. MAB3232 reacts with basal cell compartments of stratified and combined epithelia.Reacts with cytokeratin 14, expressed in the basal cells of the larynx, esophagus, trachea, bladder, prostate, cervix, vagina, breast acini, skin and sweat glands.

Immunogen

Cytoskeletal preparation of TR146 epithelial cells.

Application

Detect Cytokeratin 14 using this Anti-Cytokeratin 14 Antibody, clone RCK107 validated for use in FC, WB, IC, IH.
Research Category
Cell Structure
Research Sub Category
Cytokeratins
Western blot

Immunohistochemistry on frozen tissue sections.

Immunocytochemistry

Flow cytometry

Optimal working dilutions must be determined by the end user.

Physical form

Format: Purified
Liquid in buffer with 0.1% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots up to 6 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sicong Jiang et al.
Molecular medicine reports, 14(3), 1891-1900 (2016-07-20)
The anterograde intraflagellar transport motor protein, kif3a, regulates the integrity of primary cilia and various cellular functions, however, the role of kif3a in dental mesenchymal stem/precursor cell differentiation remains to be fully elucidated. In the present study, the expression of
Ziyue Li et al.
Cell proliferation, 50(4) (2017-07-18)
Schwann cells (SCs) are the principal glial cells in peripheral nerve system, involved in neuropathies with great regenerative potential. Dental pulp cells have been reported to maintain neurogenic potential. In contrast, the regulatory role of SCs on human dental pulp
Inflammatory marker analysis in psoriatic skin under topical phosphodiesterase 4 inhibitor treatment.
Lennart M Roesner et al.
The Journal of allergy and clinical immunology, 140(4), 1184-1187 (2017-05-17)
Kai-Chiang Yang et al.
Journal of tissue engineering and regenerative medicine, 6(10), 777-785 (2011-10-29)
Odontogenesis is a complex process with a series of epithelial-mesenchymal interactions and odontogenic molecular cascades. In tissue engineering of teeth from stem cells, platelet-rich fibrin (PRF), which is rich in growth factors and cytokines, may improve regeneration. Accordingly, PRF was
Jiawei Zhang et al.
Journal of cellular physiology, 234(7), 11900-11911 (2018-12-26)
Dental follicle cells (DFCs) activate and recruit osteoclasts for tooth development and tooth eruption, whereas DFCs themselves differentiate into osteoblasts to form alveolar bone surrounding tooth roots through the interaction with Hertwig's epithelial root sheath (HERS). Also during tooth development

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service