Skip to Content
Merck
All Photos(1)

Key Documents

MAB3194

Sigma-Aldrich

Anti-Glutathione: N-ethylmaleimide adduct Antibody, clone 8.1GSH

clone 8.1GSH, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

8.1GSH, monoclonal

species reactivity (predicted by homology)

mammals

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

Specificity

Reactive with thiol-modified glutatione. Does not react with oxidized glutathione (GSSG) or glutathione (GSH) in mixed disulfide linkage with proteins. MAB3194 is specific to the glutathione:N-ethylmaleimide adduct; thus it is useful for quantification of reduced glutathione (GSH) after reaction with N-ethylmaleimide. For use in ELISA or flow cytometry, sample must first be reacted with NEM.

Immunogen

GS-NEM conjugated to KLH

Application

Detect the Glutathione: N-ethylmaleimide adduct protein using this Anti-Glutathione: N-ethylmaleimide adduct, clone 8.1GSH validated for use in ELISA, Flow, ICC, IHC & WB.
Immunohistochemistry: 1:100 (paraformaldehyde fixation)

Immunocytochemistry: 1:100 - 1:500 for assessment of intracellular GSH distribution.

ELISA: 1:100,000 - 1:200,000 to quantify GSH at >2 mM in biological specimens.

Flow cytometry: 1:100 - 1:1,000

Cells were fixed with cold 1% PFA for 5 min and vortexed into methanol (-20°C) containing 5mM NEM (10 min on dry ice) or 10 μM NEM (N-ethyl maleimide) for 30 min at RT.

Optimal working dilutions must be determined by end user.

Positive Control: Peripheral blood mononuclear cells (PBMC) treated with NEM
Research Category
Neuroscience
Research Sub Category
Oxidative Stress

Physical form

Format: Purified
Liquid in 0.02M PBS pH 7.6, 0.25M NaCl containing 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad Nizam Uddin et al.
Cell stress & chaperones, 28(2), 201-217 (2023-02-17)
Oxidative stress including decreased antioxidant enzyme activities, elevated lipid peroxidation, and accumulation of advanced glycation end products in the blood from children with autism spectrum disorders (ASD) has been reported. The mechanisms affecting the development of ASD remain unclear; however
Gobinath Shanmugam et al.
Frontiers in physiology, 8, 268-268 (2017-05-19)
Nuclear factor erythroid 2 related factor 2 (Nrf2) signaling maintains the redox homeostasis and its activation is shown to suppress cardiac maladaptation. Earlier we reported that acute endurance exercise (2 days) evoked antioxidant cytoprotection in young WT animals but not
Carole Escartin et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 31(20), 7392-7401 (2011-05-20)
Astrocytes support neuronal antioxidant capacity by releasing glutathione, which is cleaved to cysteine in brain extracellular space. Free cysteine is then taken up by neurons through excitatory amino acid transporter 3 [EAAT3; also termed Slc1a1 (solute carrier family 1 member
Reiko Satow et al.
Gastroenterology, 142(3), 572-581 (2011-12-14)
Loss of promyelocytic leukemia protein (PML) nuclear body (NB) formation has been reported in colorectal and other solid tumors. However, genetic alteration of PML is rarely observed in these tumors; the exact mechanisms that mediate loss of PML function are
Radhakrishnan Rajesh Kumar et al.
Journal of translational medicine, 14, 86-86 (2016-04-07)
Anomalies in myocardial structure involving myocyte growth, hypertrophy, differentiation, apoptosis, necrosis etc. affects its function and render cardiac tissue more vulnerable to the development of heart failure. Although oxidative stress has a well-established role in cardiac remodeling and dysfunction, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service