Skip to Content
Merck
All Photos(1)

Key Documents

MAB2636

Sigma-Aldrich

Anti-SERCA2 Antibody, clone IID8

clone IID8, from mouse

Synonym(s):

Sarcoplasmic/endoplasmic reticulum calcium ATPase 2, SR Ca(2+)-ATPase 2, ATPase, Ca++ transporting, cardiac muscle, slow twitch 2, Calcium pump 2, Calcium-transporting ATPase sarcoplasmic reticulum type, slow twitch skeletal muscle isoform, Endoplasmic r

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

IID8, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ATP2A2(488)

General description

The sarcoplasmic reticulum of striated muscle is a specialized intracellular membrane system that plays a critical role in the contraction and relaxation of muscle. The sarco/endoplasmic reticulum Ca2+ ATPases (SERCAs) mediate Ca2+ uptake into intracellular stores. SERCA mediated Ca2+ uptake induces and maintains muscular relaxation. SERCA1 is expressed in fast-twitch muscles, whereas SERCA2 is expressed in cardiac and slow-twitch muscles. SERCA2 plays an important part in regulating cardiac contractile function.

Specificity

The antibody recognizes SERCA2.

Immunogen

Epitope: Unknown
Purified canine cardiac sarcoplasmic reticulum.

Application

Anti-SERCA2 Antibody, clone IID8 is an antibody against SERCA2 for use in WB, IC, IH.
Immunocytochemistry Analysis: A previous lot was used by an independent laboratory in IC (Eizema, 2007).
Research Category
Cell Structure
Research Sub Category
Cytoskeleton

Cytoskeletal Signaling

Quality

Evaluated by Western Blot in HEK293 cell lysate.

Western Blot Analysis: 0.5 µg/ml of the antibody detected SERCA2 in 10 µg of HEK293 cell lysate.

Target description

~ 110 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HEK293 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akihiro Yoneda et al.
Cancer science, 112(7), 2803-2820 (2021-06-11)
Pancreatic ductal adenocarcinoma (PDAC) is one of the most chemoresistant cancers. An understanding of the molecular mechanism by which PDAC cells have a high chemoresistant potential is important for improvement of the poor prognosis of patients with PDAC. Here we
Tao Liu et al.
JCI insight, 5(8) (2020-04-08)
Atrial fibrillation (AF) alters atrial cardiomyocyte (ACM) Ca2+ handling, promoting ectopic beat formation. We examined the effects of AF-associated remodeling on Ca2+-related action potential dynamics and consequences for AF susceptibility. AF was maintained electrically in dogs by right atrial (RA)
Mirella Telles Salgueiro Barboni et al.
Progress in retinal and eye research, 95, 101137-101137 (2022-11-21)
Duchenne muscular dystrophy (DMD) is caused by X-linked inherited or de novo DMD gene mutations predominantly affecting males who develop early-onset muscle degeneration, severely affecting their quality of life and leading to reduced life expectancy. DMD patients may also develop
Shivam A Zaver et al.
JCI insight, 8(18) (2023-08-10)
Mutation of the ATP2A2 gene encoding sarco-endoplasmic reticulum calcium ATPase 2 (SERCA2) was linked to Darier disease more than 2 decades ago; however, there remain no targeted therapies for this disorder causing recurrent skin blistering and infections. Since Atp2a2-knockout mice
Tomohisa Seki et al.
PloS one, 9(1), e85645-e85645 (2014-01-28)
Induced pluripotent stem cells (iPSCs) have been proposed as novel cell sources for genetic disease models and revolutionary clinical therapies. Accordingly, human iPSC-derived cardiomyocytes are potential cell sources for cardiomyocyte transplantation therapy. We previously developed a novel generation method for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service