Skip to Content
Merck
All Photos(1)

Key Documents

MAB2627

Sigma-Aldrich

Anti-VCAM-1 (CD106) Antibody, clone 6C7.1

clone 6C7.1, from rat

Synonym(s):

vascular cell adhesion molecule 1, vascular cell adhesion protein 1, CD106 antigen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6C7.1, monoclonal

species reactivity

mouse

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

Related Categories

General description

Vascular cell adhesion protein 1 (VCAM-1) is a cell surface adhesion molecule belonging to the immunoglobulin cell-cell adhesion receptor family and is comprised of seven Ig-like C2-type domains. VCAM-1 is a crucial component in cell-cell recognition and seems to be involved in cellular adhesion between endothelial cells and leukocytes. It has a regulatory role for both signal transduction and adhesion through its interaction with leukocyte beta-1 integrin VLA4, an interaction that is potentially involved in immune and inflammatory emigration responses of leukocytes. Studies have shown upregulation of VCAM-1 to be critical in responding to inflammatory diseases such as atherosclerosis. VCAM-1 is primarily expressed in endothelial cells and is expressed as two known isoforms found within the cell membrane. Isoform 1 functions as a single-pass type 1 membrane protein, while Isoform 2 acts as a GPI-anchor.

Immunogen

Mouse endothelial cells

Application

Anti-VCAM-1 (CD106) Antibody, clone 6C7.1 is an antibody against VCAM-1 (CD106) for use in FC, IP, IC, IH.
Immunohistochemistry Analysis: A previous lot was used by independent laboratory in IH. (Norman, U., et al. (2008). Journal of Leukocyte Biology. 84:68-76.)

Immunocytochemistry Analysis: A previous lot was used by independent laboratory in IC. (Norman, U., et al. (2008). Journal of Leukocyte Biology. 84:68-76.)

Quality

Evaluated by Flow Cytometry in mouse bone marrow cells.

Flow Cytometry Analysis: 10 µg of this antibody detected VCAM-1 on mouse bone marrow cells.

Target description

81 kDa calculated

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David S Ojala et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 26(1), 304-319 (2017-10-11)
Directed evolution continues to expand the capabilities of complex biomolecules for a range of applications, such as adeno-associated virus vectors for gene therapy; however, advances in library design and selection strategies are key to develop variants that overcome barriers to
Gahee Min et al.
Journal of cellular and molecular medicine, 20(12), 2333-2340 (2016-07-16)
Drug repositioning is used to discover drug candidates to treat human diseases, through the application of drugs or compounds that are approved for the treatment of other diseases. This method can significantly reduce the time required and cost of discovering
Chao Ma et al.
Science advances, 6(44) (2020-11-01)
B cell acute lymphoblastic leukemia (B-ALL) blasts hijack the bone marrow (BM) microenvironment to form chemoprotective leukemic BM "niches," facilitating chemoresistance and, ultimately, disease relapse. However, the ability to dissect these evolving, heterogeneous interactions among distinct B-ALL subtypes and their
Peter Baluk et al.
Methods in molecular biology (Clifton, N.J.), 2441, 115-134 (2022-02-01)
Changes in blood vessels and lymphatics in health and disease are easier to understand and interpret when studied microscopically in three dimensions. The mouse trachea is a simple, yet powerful, and versatile model system in which to achieve this. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service