Skip to Content
Merck
All Photos(1)

Key Documents

MAB1978

Sigma-Aldrich

Anti-Integrin αV (ITGAV/CD51) Antibody

CHEMICON®, mouse monoclonal, LM142

Synonym(s):

CD51

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Integrin αV Antibody, clone LM142, ascites fluid, clone LM142, Chemicon®

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

LM142, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1

suitability

not suitable for activity/function inhibition

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ITGAV(3685)

Specificity

Reactive with integrin alpha-V.

Application

Detect Integrin αV using this Anti-Integrin αV Antibody, clone LM142 validated for use in IP, IH.
Immunohistochemistry (Acetone Fixed Only) 1:1000

Immunofluorescence and immunoprecipitation.

MAB1978 is non-blocking.

Final working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Integrins

Storage and Stability

Maintain at -20°C for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatiana Gianni et al.
PLoS pathogens, 9(12), e1003806-e1003806 (2013-12-25)
Herpes simplex virus (HSV)--and herpesviruses in general--encode for a multipartite entry/fusion apparatus. In HSV it consists of the HSV-specific glycoprotein D (gD), and three additional glycoproteins, gH/gL and gB, conserved across the Herpesviridae family and responsible for the execution of
M O De Nichilo et al.
The Journal of biological chemistry, 271(18), 11016-11022 (1996-05-03)
The macrophage colony-stimulating factor (M-CSF) is able to induce the expression of the alpha v beta 5 integrin receptor on the surface of cultured human macrophages (De Nichilo, M. O., and Burns, G. F. (1993) Proc. Natl. Acad. Sci. U.S.A.
Masako Ueda et al.
Biomaterials, 31(25), 6394-6399 (2010-06-12)
We previously showed that blood outgrowth endothelial cells (BOECs) had a high affinity for polyurethane (PU) covalently configured with cholesterol residues (PU-Chol). However, the molecular mechanisms responsible for this enhanced affinity were not determined. CD47, a multifunctional transmembrane glycoprotein involved
Tobias G Kapp et al.
Scientific reports, 7, 39805-39805 (2017-01-12)
Integrins, a diverse class of heterodimeric cell surface receptors, are key regulators of cell structure and behaviour, affecting cell morphology, proliferation, survival and differentiation. Consequently, mutations in specific integrins, or their deregulated expression, are associated with a variety of diseases.
Robert McMillan et al.
British journal of haematology, 118(4), 1132-1136 (2002-08-30)
In chronic immune thrombocytopenic purpura (ITP), autoantibodies bind to platelet surface proteins, particularly alphaIIb, resulting in platelet destruction by the reticulo-endothelial system. In order to better localize the autoepitopes on alphaIIb, we studied the binding of antibodies to Chinese hamster

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service