Skip to Content
Merck
All Photos(1)

Key Documents

MAB1621

Sigma-Aldrich

Anti-Neurofilament 145 kDa Antibody, CT, clone 3H11

clone 3H11, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

3H11, monoclonal

purified by

affinity chromatography

species reactivity

avian

species reactivity (predicted by homology)

mammals

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... NEFM(4741)

General description

Neurofilaments are a type of intermediate filament that serve as major elements of the cytoskeleton supporting the axon cytoplasm. They are the most abundant fibrillar components of the axon, being on average 3-10 times more frequent than axonal microtubules. Neurofilaments (10nm in dia.) are built from three intertwined protofibrils which are themselves composed of two tetrameric protofilament complexs of monomeric proteins. The neurofilament triplet proteins (68/70, 160, and 200 kDa) occur in both the central and peripheral nervous system and are usually neuron specific. The 68/70 kDa NF-L protein can self-assemble into a filamentous structure, however the 160 kDa NF-M and 200 kDa NF-H proteins require the presence of the 68/70 kDa NF-L protein to co-assemble. Neuromas, ganglioneuromas, gangliogliomas, ganglioneuroblastomas and neuroblastomas stain positively for neurofilaments. Although typically restricted to neurons, neurofilaments have been detected in paragangliomas and adrenal and extra-adrenal pheochromocytomas. Carcinoids, neuroendocrine carcinomas of the skin, and oat cell carcinomas of the lung also express neurofilaments. For more neurofilament information see Nervous System Cell Type Specific Marker chart online under the CHEMICON Technical Support section.

Specificity

Reacts with an epitope dependent on the last 84 amino acids of rat NF-M (145K). Reacts specifically with NF-M from a variety of mammalian and avian species. The antibody is not sensitive to the phosphorylation state of the neurofilament.

Immunogen

Recombinant fusion protein containing the last 168 amino acids of rat NF-M. Purified from E. Coli inclusion bodies by ion exchange chromatography on DEAE-cellulose in 6 M urea.

Application

Anti-Neurofilament 145 kDa Antibody, C-terminus, clone 3H11 detects level of Neurofilament 145 kDa & has been published & validated for use in WB, IC, IH.
Immunohistochemistry: 1:50-1:1000; fresh frozen, 4% PFA fixed tissue sections

Immunoblotting: 1:1000 - 1:2000 ECL detection.

Optimal working dilutions must be determined by end user.

Physical form

Affinity purified antibody in PBS, 50% glycerol, and 5 mM sodium azide

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Involvement of TRPV2 activation in intestinal movement through nitric oxide production in mice.
Mihara, H; Boudaka, A; Shibasaki, K; Yamanaka, A; Sugiyama, T; Tominaga, M
The Journal of Neuroscience null
Pierre-Yves Lozach et al.
The Journal of biological chemistry, 279(31), 32035-32045 (2004-05-29)
The molecular mechanisms involved in the hepatic tropism of hepatitis C virus (HCV) have not been identified. We have shown previously that liver-expressed C-type lectins L-SIGN and DC-SIGN bind the HCV E2 glycoprotein with high affinity (Lozach, P. Y., Lortat-Jacob
Polysaccharide-modified scaffolds for controlled lentivirus delivery in vitro and after spinal cord injury.
Thomas, AM; Shea, LD
Journal of Controlled Release : Official Journal of the Controlled Release Society null
Robin M Perelli et al.
Development (Cambridge, England), 148(9) (2021-05-08)
Angiogenesis in the developing mammalian retina requires patterning cues from astrocytes. Developmental disorders of retinal vasculature, such as retinopathy of prematurity (ROP), involve arrest or mispatterning of angiogenesis. Whether these vascular pathologies involve astrocyte dysfunction remains untested. Here, we demonstrate
Chi-Yuan Chiu et al.
European journal of pain (London, England), 26(10), 2238-2256 (2022-09-14)
Kv4 channels are key components controlling neuronal excitability at membrane potentials below action potential thresholds. It remains elusive whether Kv4.1 participates in pain regulation. We raised a Kv4.1 antibody to map Kv4.1+ neurons in the superficial dorsal horn of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service