Skip to Content
Merck
All Photos(1)

Key Documents

MAB1611

Sigma-Aldrich

Anti-Keratin Epithelial Antibody, clone AE3

clone AE3, Chemicon®, from mouse

Synonym(s):

Type II Cytokeratin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

clone

AE3, monoclonal

species reactivity

rat, chicken, rabbit, mouse, human, monkey, bovine

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Specificity

Recognizes most of the basic (Type II) keratins, HMW cytokeratins. It is broadly reactive and stains positively almost all epithelia and their neoplasms. AE3 reacts with all layers of epidermis (basal layer and above). The antibody is an excellent marker for distinguishing carcinomas from non-epithelial tumours. It has been used for studying the immunolocalisation of epidermal keratins, tissue distribution of keratins, various diseases of epidermis, oral mucosa and other epithelia and other epithelia development. AE3 and AE1 (Catalog Number MAB1612) have been used extensively for distinguishing carcinoma from non-carcinoma.

Immunogen

Human epidermal cells

Application

Anti-Keratin Epithelial Antibody, clone AE3 is an antibody against Keratin Epithelial for use in WB, IH(P).
Immunohistochemistry on unfixed frozen sections or formalin-fixed paraffin-embedded tissue sections. For paraffin-embedded sections, antigen retrieval (HEIR or trypsin treatment) is strongly recommended.

Western blotting: Clone AE3 recognizes the 65 to 67, 64, 59, 58, 56 and 52 kDa human cytokeratins of the basic subfamily.

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Cytokeratins

Linkage

Replaces: 04-588

Physical form

Format: Purified
Liquid in PBS. Contains 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
POSITIVE CONTROL: MCF-7, RT112, and A431 cell lines.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pathogenesis of fulminant monkeypox with bacterial sepsis after experimental infection with West African monkeypox virus in a cynomolgus monkey.
Nagata, N; Saijo, M; Kataoka, M; Ami, Y; Suzaki, Y; Sato, Y; Iwata-Yoshikawa, N; Ogata et al.
International Journal of Clinical and Experimental Pathology null
H Battifora et al.
Cancer, 54(5), 843-848 (1984-09-01)
The authors investigated the presence and distribution of keratin in germ cell tumors using a rabbit-anti-keratin antiserum and a monoclonal antikeratin antibody--which is specific for keratin classes of 40, 50, and 56.5 kdaltons--by various immunohistochemical methods on frozen sections, alcohol-fixed
S R Shi et al.
The American journal of pathology, 117(1), 53-63 (1984-10-01)
Nasopharyngeal carcinoma (NPC) provides a unique opportunity to evaluate distinctive epidemiologic features and a possible etiologic relationship with Epstein-Barr virus (EBV) in human malignancy. The lack of a uniformly accepted pathologic classification for NPC has limited the application of this
Viraf Sam Vasania et al.
Journal of ophthalmic & vision research, 9(4), 407-416 (2015-02-25)
To establish the efficacy and safety of ex vivo cultured autologous human conjunctival epithelial cell (hCjEC) transplantation for treatment of pterygia. Twenty-five patients with pterygia were recruited at different centers across the country. Autologous hCjEC grafts were prepared from conjunctival
Wild-type measles virus with the hemagglutinin protein of the edmonston vaccine strain retains wild-type tropism in macaques.
Takeuchi, K; Nagata, N; Kato, SI; Ami, Y; Suzaki, Y; Suzuki, T; Sato et al.
Journal of virology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service