Skip to Content
Merck
All Photos(1)

Key Documents

MAB1435

Sigma-Aldrich

Anti-Macrophages/Monocytes Antibody, clone ED-1

clone ED-1, Chemicon®, from mouse

Synonym(s):

Anti-Monocyte Antibody, Clone ED-1 Anti-Macrophages, Monocytes Detection Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

ED-1, monoclonal

species reactivity

rat

should not react with

horse

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
radioimmunoassay: suitable
western blot: suitable

isotype

IgG1

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

General description

Monocytes are produced by the bone marrow from haematopoietic stem cell precursors called monoblasts. Monocytes circulate in the bloodstream for about one to three days and then typically move into tissues throughout the body. They consist of between 3 to 8 percent of the leukocytes in the blood. In the tissues monocytes mature into different types of macrophages at different anatomical locations.
Macrophages are a type of white blood cell derived from monocytes that engulf invading antigenic molecules, viruses, and microorganisms and then display fragments of the antigen to activate helper T cells; ultimately stimulating the production of antibodies against the antigen.

Specificity

Recognizes a single chain glycoprotein of 90-100 kD that is expressed on the lysomal membrane of myeloid cells. Weak cell surface expression also occurs. The antigen is expressed by the majority of tissue macrophages and weakly by peripheral blood granulocytes. Studies have shown that the antigen has many characteristics in common with macrosialin in the mouse and CD68 in the human.
Clone ED1, which recognises a CD68 like molecule in rat is an excellent marker of activated microglia. For pan microglial markers, staining both resting and activated microglia, use like CD11b (MAB1387Z and others) or iba-1 (MABN92).

Immunogen

Rat spleen cells

Application

Immunohistology on frozen sections at 1:50-1:300. Paraffin embedded tissue (Bouin fixation): 1:50-1:300 (requires pretreatment with trypsin or pronase).

Flow cytometry at 1:100 - 1:200. Use 10 μL of this working dilution to label 10E6 cells.

Radioimmunoassay

Also suitable for Immunoprecipitation, and Western Blotting.

Optimal working dilutions must be determined by end user.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This Anti-Macrophages/Monocytes Antibody, clone ED-1 is validated for use in Flow Cytometry (FC), Immunohistochemistry (IHC), Immunoprecipitation (IP), Radioimmunoassay (RIA), and Western Blotting (WB) for the detection of CD68.

Target description

90-100 kD

Linkage

Replaces: CBL1511; CBL260

Physical form

Format: Purified
Protein A purified.
Purified mouse IgG1 in PBS (pH 7.4) and 0.09% sodium azide

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Macrophages, monocytes, lymphoid organs

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Kohki Matsubara et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(6), 2452-2464 (2015-02-13)
Engrafted mesenchymal stem cells from human deciduous dental pulp (SHEDs) support recovery from neural insults via paracrine mechanisms that are poorly understood. Here we show that the conditioned serum-free medium (CM) from SHEDs, administered intrathecally into rat injured spinal cord
L Yao et al.
Gene therapy, 20(12), 1149-1157 (2013-07-26)
Functionalized biomaterial scaffolds targeted at improving axonal regeneration by enhancing guided axonal growth provide a promising approach for the repair of spinal cord injury. Collagen neural conduits provide structural guidance for neural tissue regeneration, and in this study it is
Daniel L Pouliquen et al.
Oncotarget, 8(34), 57552-57573 (2017-09-17)
A rat model of sarcomatoid mesothelioma, mimicking some of the worst clinical conditions encountered, was established to evaluate the therapeutic potential of intracavitary curcumin administration. The M5-T1 cell line, selected from a collection established from F344 rats induced with asbestos
Y Suzuki et al.
Biotechnic & histochemistry : official publication of the Biological Stain Commission, 87(4), 241-248 (2011-10-01)
The effects of prolonged water washing after fixation for 48 h in 10% (v/v) phosphate-buffered neutral formalin on the quality of representative histological staining methods were evaluated using samples of liver, kidney, spleen and thymus collected from three male Crl:CD(SD)(IGS)
Kendall Mitchell et al.
BMC neuroscience, 17(1), 80-80 (2016-12-03)
Bone morphogenetic protein-2 (BMP-2) is a pleiotropic, secreted molecule with diverse effects. The potent ability of BMP-2 to stimulate bone growth prompted its widespread clinical use for arthrodesis (spine fusion). However, elevated post-operative pain in patients treated with BMP-2 has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service