Skip to Content
Merck
All Photos(1)

Key Documents

MAB13424

Sigma-Aldrich

Anti-MMP-13 Antibody, clone VIIIA2

clone VIIIA2, Chemicon®, from mouse

Synonym(s):

Collagenase-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

VIIIA2, monoclonal

species reactivity

mouse, human, rabbit

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MMP13(4322)

Specificity

It recognizes proteins of ~60 kDa and ~48 kDa which are identified as pro (latent) and activate forms of matrix metalloproteinase-13 (MMP-13; also known as Collagenase-3). Shows no cross reactivity with the pro and active forms of other MMPs. Human collagenase-3 (MMP13) is a recently identified member of the matrix metalloproteinase (MMP) family that is expressed in breast carcinomas and in articular cartilage from arthritic patients. The MMP-13 gene has been isolated and characterized. This gene is composed of 10 exons and 9 introns and spans over 12.5 kb. The overall organization of the collagenase-3 gene is similar to that of other MMP genes clustered at chromosome 11q22, including fibroblast collagenase (MMP-1), matrilysin (MMP-7), and macrophage metalloelastase (MMP-12), but is more distantly related to genes coding for stromelysin-3 (MMP-11), gelatinase-A (MMP-2), and gelatinase-B (MMP-9), which map outside of this gene cluster. Cellular Localization: Cytoplasmic

Immunogen

Human recombinant collagenase-3 protein.

Application

Immunohistochemistry (frozen and formalin-fixed/paraffin*): 2-4 μg/mL for 60 minutes at room temperature.

* Staining of formalin-fixed tissues is enhanced by boiling tissue sections in 10 mM citrate buffer, pH 6.0, for 10-20 minutes followed by cooling at room temperature for 20 minutes.

Immunoprecipation: use protein G (2 μg/mg protein lysate)

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
MMPs & TIMPs
This Anti-MMP-13 Antibody, clone VIIIA2 is validated for use in IH for the detection of MMP-13.

Physical form

Format: Purified
Liquid in 10 mM PBS, pH 7.4, with 0.2% BSA and 15 mM sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 12 months.

Analysis Note

Control
POSITIVE CONTROL: Conditioned, serum-free medium from (PMA-treated) human fibrosarcoma HT-1080 cells. Bladder, breast, ovarian carcinomas.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bu-Nam Jeon et al.
FEBS letters, 593(18), 2665-2674 (2019-06-22)
Dysregulated matrix metalloproteinase (MMP) gene expression is a major cause of the degradation of lung tissue that is integral to emphysema pathogenesis. Cigarette smoking (CS) increases MMP gene expression, a major contributor to emphysema development. We previously reported that Zbtb7c
Ann M Kennedy et al.
The Journal of clinical investigation, 115(10), 2832-2842 (2005-09-17)
MMPs, which degrade components of the ECM, have roles in embryonic development, tissue repair, cancer, arthritis, and cardiovascular disease. We show that a missense mutation of MMP13 causes the Missouri type of human spondyloepimetaphyseal dysplasia (SEMD(MO)), an autosomal dominant disorder
Kaige Ma et al.
Science advances, 10(7), eadi5501-eadi5501 (2024-02-14)
Osteoarthritis (OA) is characterized by cartilage damage, inflammation, and pain. Vascular endothelial growth factor receptors (VEGFRs) have been associated with OA severity, suggesting that inhibitors targeting these receptors alleviate pain (via VEGFR1) or cartilage degeneration (via VEGFR2). We have developed
Hongyuan Zhang et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 37(5), 983-996 (2022-02-28)
Enchondromas and chondrosarcomas are common cartilage neoplasms that are either benign or malignant, respectively. The majority of these tumors harbor mutations in either IDH1 or IDH2. Glutamine metabolism has been implicated as a critical regulator of tumors with IDH mutations.
Pegah Rahmati Nezhad et al.
Experimental dermatology, 30(11), 1631-1641 (2021-04-05)
The incidence of cutaneous squamous cell carcinoma (cSCC) is increasing globally. Here, we have studied the functional role of complement factor I (CFI) in the progression of cSCC. CFI was knocked down in cSCC cells, and RNA-seq analysis was performed.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service