Skip to Content
Merck
All Photos(3)

Key Documents

LUXRGTKIT

Millipore

mPAGE® Lux Bis-Tris Reagent Kit

greener alternative

Reagent Kit of Resolving Solution, Diluent and Stacking Solution for MPAGE® Lux Curing Station

Synonym(s):

mPAGE® Lux Bis-Tris Reagent Kit for mPAGE® Lux Curing Station

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105323
NACRES:
NC.01

product name

mPAGE® Lux Bis-Tris Reagent Kit,

shelf life

12 mo.

Quality Level

packaging

kit of 1 kit (~75 gels)

manufacturer/tradename

Millipore

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

SDS-PAGE: suitable
electrophoresis: suitable
immunoblotting: suitable
western blot: suitable

greener alternative category

storage temp.

2-8°C

General description

The mPAGE® Lux Bis-Tris Reagent Kit is the only reagent kit which is compatible with mPAGE® Lux Curing Station for making polyacrylamide gels. It offers rapid and simple gel curing process for handcasting mini protein gels. A mini gel is cured in 1.5min and overall casting process can be less than 3min. It is a kit of mPAGE® Lux Resolving Solution (225 mL), mPAGE® Lux Diluent (225 mL) and mPAGE® Lux Stacking Solution (112 mL).
We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives . This product is a Design for Sustainability (DfS) developed product. Click here to view its DfS scorecard.

Components

The mPAGE® Lux Bis-Tris Reagent Kit is a kit includes the following components:
  • mPAGE® Lux Bis-Tris Resolving Solution
  • mPAGE® Lux Bis-Tris Diluent
  • mPAGE® Lux Bis-Tris Stacking Solution

Compatibility

Compatible with:
  • mPAGE® Lux Curing Station
  • mPAGE® Lux Masked Short Plates
  • mPAGE® Lux Mixing Tubes
  • mPAGE® Combs
  • mPAGE® Mini Spacer Plates
  • mPAGE® Mini Gel Tank
  • mPAGE® Mini Wet Transfer System

Target description

The mPAGE® Lux Bis-Tris Reagent Kit is a kit includes the following components:
  • mPAGE® Lux Bis-Tris Resolving Solution
  • mPAGE® Lux Bis-Tris Diluent
  • mPAGE® Lux Bis-Tris Stacking Solution

Legal Information

MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Refresh your gel casting workflow with ready-to-use SDS-PAGE gels in just 3 minutes. Discover faster, simpler, and safer gel casting methods.

Refresh your gel casting workflow with ready-to-use SDS-PAGE gels in just 3 minutes. Discover faster, simpler, and safer gel casting methods.

Refresh your gel casting workflow with ready-to-use SDS-PAGE gels in just 3 minutes. Discover faster, simpler, and safer gel casting methods.

Refresh your gel casting workflow with ready-to-use SDS-PAGE gels in just 3 minutes. Discover faster, simpler, and safer gel casting methods.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service