JU-125ML-BK
Intermediates For Further Manufacturing Use (FFMU) - Anti Human Globulin
Anti-C3d from the clone BRIC-8, Murine IgM, Intermediate for further manufacturing use
Synonym(s):
Anti-C3d (clone BRIC-8), FFMU
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
unpurified
clone
BRIC-8, monoclonal
form
liquid
packaging
pkg of 125 mL
concentration
2-10 mg/mL protein (Total Protein Range)
technique(s)
agglutination assay: suitable
pH
6.7-7.7
isotype
IgM
shipped in
ambient
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
For the formulation of Anti-Human Globulin blood typing reagents or devices.
Contains 0.1% (w/v) Sodium Azide; Shelf Life: 24 months from date of filtration.
Licensed by the US FDA under a shared manufacturing agreement.
Application
Legal Information
Disclaimer
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service