Skip to Content
Merck
All Photos(1)

Key Documents

ISEQ85R

Millipore

Immobilon®-PSQ Membrane

8.5 cm x 10 m roll, 0.2 µm pore size, Hydrophobic PVDF Transfer Membrane

Synonym(s):

Immobilon® NOW, Immobilon® narrow roll, Western blotting membrane, blotting membrane, transfer membrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105339
eCl@ss:
32031613
NACRES:
NB.23
Pricing and availability is not currently available.

Product Name

Immobilon®-PSQ Membrane, PVDF, 0.2 m, 8.5 cm x 10 m roll, 0.2 um filter ( 0.2 micron ) pore size Hydrophobic PVDF Transfer Membrane for western blotting using Chemiluminescence, Chromogenic or Radioactive detection methods. Comes in an 8.5 cm x 10 m roll.

material

PVDF membrane
plain filter
white filter

Quality Level

feature

hydrophobic

manufacturer/tradename

Immobilon®

technique(s)

dot blot: suitable
western blot: suitable

filter L × W

8.5 cm × 10 m

pore size

0.2 μm pore size

capacity

262 μg/cm2 adsorption capacity (insulin)
340 μg/cm2 adsorption capacity (BSA)
448 μg/cm2 adsorption capacity (goat IgG)

compatibility

for use with Amido black
for use with CPTS
for use with Colloidal gold
for use with Coomassie brilliant blue
for use with India ink
for use with Ponceau-S red
for use with Sypro<TMSYMBOL></TMSYMBOL> ruby
for use with Toluidine blue
for use with Transillumination

detection method

chemiluminescent
colorimetric
radioactive

shipped in

ambient

General description

The Immobilon®-PSQ membrane is a 0.2 μm microporous polyvinylidene fluoride (PVDF) membrane. It is for protein sequencing and immunodetection of proteins with molecular weights of less than 20,000 Daltons.
Filter Code: ISEQ
Primary Binding Mechanisms: Electrostatic, hydrophobic

Application

Immobilon®-PSQ Membrane, PVDF, 0.2 μm, 8.5 cm x 10 m roll has been used in western blot analysis.[1][2][3]

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuelai Feng et al.
Biomedical reports, 13(2), 6-6 (2020-07-02)
The effect of Apocynum venetum leaf extract (AVLE) on the nervous system has been widely studied, but its effect on injured neurons is not fully understood. In the present study, the protective effect of AVLE on injured neurons was determined.
Dattatraya Gautam Rawale et al.
Chemical science, 12(19), 6732-6736 (2021-05-28)
The conservation of chemoselectivity becomes invalid for multiple electrophilic warheads during protein bioconjugation. Consequently, it leads to unpredictable heterogeneous labeling of proteins. Here, we report that a linchpin can create a unique chemical space to enable site-selectivity for histidine and
Evangelia Sereti et al.
Biomedicines, 9(1) (2021-01-21)
Sigma (σ) receptors have attracted great interest since they are implicated in various cellular functions and biological processes and diseases, including various types of cancer. The receptor family consists of two subtypes: sigma-1 (σ1) and sigma-2 (σ2). Both σ receptor

Articles

A discussion of factors affecting observed vs. calculated molecular weight in gel electrophoresis and western blotting.

Protocols

This page shows and discusses three protocols for stripping and reprobing a western blot membrane.

Simplify your SDS-PAGE gel preparation with our pre-mixed solutions and protocols to achieve precise protein separation.

Simplify your SDS-PAGE gel preparation with our pre-mixed solutions and protocols to achieve precise protein separation.

Simplify your SDS-PAGE gel preparation with our pre-mixed solutions and protocols to achieve precise protein separation.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service