Skip to Content
Merck
All Photos(1)

Key Documents

IPFL07810

Millipore

Immobilon®-FL PVDF Membrane

1 roll, 7 x 8.4cm, 0.45 µm pore size, Hydrophobic PVDF Transfer Membrane with low background fluorescence for Western blotting. Compatible with visible and infrared fluorescent probes.

Synonym(s):

Western blotting membrane, blotting membrane, transfer membrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105339
eCl@ss:
32031602
NACRES:
NB.22

product name

Immobilon®-FL PVDF Transfer Membrane, 0.45 μm pore size, 7 X 8.4 cm sheets

material

PVDF membrane
plain filter
white filter

Quality Level

feature

hydrophobic

manufacturer/tradename

Immobilon®

technique(s)

dot blot: suitable
western blot: suitable

filter L × W

7 cm × 8.4 cm

pore size

0.45 μm pore size

capacity

155 μg/cm2 adsorption capacity (insulin)
205 μg/cm2 adsorption capacity (BSA)
300 μg/cm2 adsorption capacity (goat IgG)

compatibility

for use with Amido black
for use with CPTS
for use with Coomassie brilliant blue
for use with Ponceau-S red

detection method

chemiluminescent
colorimetric
fluorometric

shipped in

ambient

General description

The Immobilon®-FL transfer membrane is a polyvinylidene fluoride (PVDF) microporous membrane for binding proteins transferred from a variety of gel matrices. The Immobilon®-FL membrane has a nominal pore size of 0.45 μm and is recommended for blotting proteins with molecular weights greater than 20 kilodaltons (kDa). The membrane exhibits very low autofluorescence across a wide range of excitation/emission wavelengths. This property makes it ideal for blotting applications involving fluorescence-based immunodetection.
Filter Code: IPFL

Application

Immobilon®-FL Transfer Membrane has been used in western blotting.

Features and Benefits

Immobilon®-FL Transfer Membrane:
  • Designed for optimal fluorescent immunodetection application
  • Extremely low background improves the sensitivity of all fluorescence based immunodetection
  • Compatible with all commonly used fluorescent probes across a wide range of excitation/emission wavelengths
  • For standard chemiluminescent or chromogenic detection

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shaimaa I A Ibrahim et al.
Frontiers in cellular neuroscience, 12, 453-453 (2018-12-14)
Low back pain, a leading cause of disability, is commonly treated by epidural steroid injections that target the anti-inflammatory glucocorticoid receptor (GR). However, their efficacy has been controversial. All currently used epidural steroids also activate the pro-inflammatory mineralocorticoid receptor (MR)
Rianne N Esquivel et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 27(5), 974-985 (2019-04-10)
Zika virus (ZIKV) infection is endemic to several world regions, and many others are at high risk for seasonal outbreaks. Synthetic DNA-encoded monoclonal antibody (DMAb) is an approach that enables in vivo delivery of highly potent mAbs to control infections. We
Ryan C Bates et al.
Pharmacology, biochemistry, and behavior, 103(2), 237-244 (2012-09-11)
Valproic acid (VPA) is the most widely prescribed antiepileptic drug due to its ability to treat a broad spectrum of seizure types. However, potential complications of this drug include anticonvulsant polytherapy metabolism, organ toxicity and teratogenicity which limit its use
Kevin K Caldwell et al.
Neurotoxicology and teratology, 66, 102-112 (2017-11-15)
Our previous studies suggest that prenatal arsenic exposure (50ppb) modifies epigenetic control of the programming of the glucocorticoid receptor (GR) signaling system in the developing mouse brain. These deficits may lead to long-lasting consequences, including deficits in learning and memory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service