Skip to Content
Merck
All Photos(1)

Key Documents

IM32

Sigma-Aldrich

Anti-TIMP-1 (Ab-1) Mouse mAb (7-6C1)

liquid, clone 7-6C1, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

7-6C1, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

rat, bovine, human

should not react with

mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TIMP1(7076)

General description

Matrix metalloproteinases (MMPs) are a family of enzymes that are responsible for the degradation of extracellular matrix components such as collagen, laminin and proteoglycans. In addition to sequence homology, all MMPs share the following characteristics: the catalytic mechanism is dependent upon a zinc ion at the active center, they cleave one or more extracellular matrix components, they are secreted as zymogens which are activated by removal of an approximately 10 kDa segment from the N-terminus and their activity is regulated by endogenous inhibitors. These enzymes are involved in normal physiological processes such as embryogenesis and tissue remodeling and may play an important role in arthritis, periodontitis, and metastasis.The activation and activity of MMPs are regulated by a family of endogenous inhibitors, tissue inhibitors of metalloproteinase (TIMP). TIMP-1 (also called EPA, Fibroblast collagenase inhibitor or collagenase inhibitor) is a 28 kDa glycoprotein that is expressed by a variety of cell types. It forms a non-covalent, stoichiometric complex with both latent and active MMPs. TIMP-1 preferentially binds and inhibits MMP-9. TIMPs are capable of altering the metastatic potential of cancer cells and have been shown to inhibit invasion and metastasis in animal models.
Purified mouse monoclonal antibody (see application references). Recognizes the ~28 kDa TIMP-1 protein.
Recognizes the ~28.5 kDa TIMP-1 protein. Also recognizes MMP/TIMP-1 complexes.
This Anti-TIMP-1 (Ab-1) Mouse mAb (7-6C1) is validated for use in Immunoblotting, Immunocytochemistry, Paraffin Sections for the detection of TIMP-1 (Ab-1).

Immunogen

Bovine
purified, bovine dental pulp TIMP-1

Application


Immunoblotting (1 g/ml, see application references)
Immunocytochemistry (see application references)
Paraffin Sections (25 g/ml, see application references, see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 100 mM sodium phosphate buffer, 0.1% BSA, pH 7.0.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
rTIMP-1 Protein (Cat. Nos. PF019 or PF020)

Other Notes

Also recognizes TIMP-1/MMP complexes.This antibody has been used to stain paraffin sections fixed with PLP (perioddate-lysine-4% paraformaldehyde) and 10% formaldehyde (using 25 µg/ml antibody). Antibody should be titrated for optimal results in individual systems.
Fujimoto, N. and Iwata, K., 1994. Connective Tissue26, 237.
Cottam, D. W. and Rees, R. C., 1993. Intl. J. Oncol.2, 861.
Stetler-Stevenson, W. G., et al. 1993. FASEB J.7, 1434.
Woessner, J. F., 1991. FASEB J.5, 2145.
Liotta, L. A. and Stetler-Stevenson, W. G. 1990. in Seminars in Cancer Biology ed. M. M. Gottesman. Vol. 99-106.

Legal Information

Manufactured by Daiichi Fine Chemical Co., Ltd. Not available for sale in Japan.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S M Cork et al.
Oncogene, 31(50), 5144-5152 (2012-02-15)
Brain-specific angiogenesis inhibitor 1 (BAI1), an orphan G protein-coupled receptor-type seven transmembrane protein, was recently found mutated or silenced in multiple human cancers and can interfere with tumor growth when overexpressed. Yet, little is known about its regulation and the
Susan Notohamiprodjo et al.
Biological chemistry, 393(12), 1463-1470 (2013-05-15)
Tissue inhibitor of metalloproteinase 1 (TIMP-1) controls matrix metalloproteinase activity through 1:1stoichiometric binding. Human TIMP-1 fused to a glycosylphosphatidylinositol(GPI) anchor (TIMP-1 - GPI) shifts the activity of TIMP-1 from the extracellular matrix to the cell surface. TIMP-1 - GPI treated
Mengli Chen et al.
Journal of ethnopharmacology, 143(2), 448-454 (2012-07-17)
Cordyceps sinensis is a fungus used in traditional Chinese medicine as a tonic to soothe the lung for the treatment of fatigue and respiratory diseases. Idiopathic pulmonary fibrosis is a chronic, irreversible and debilitating lung disease showing fibroblast/myofibroblast expansion and
Georgina González-Avila et al.
Experimental and therapeutic medicine, 12(3), 1419-1427 (2016-09-08)
Asthma airway remodeling is characterized by the thickening of the basement membrane (BM) due to an increase in extracellular matrix (ECM) deposition, which contributes to the irreversibility of airflow obstruction. Interstitial collagens are the primary ECM components to be increased
Hsueh-Liang Fu et al.
The Journal of biological chemistry, 288(17), 12114-12129 (2013-03-23)
The discoidin domain receptors (DDRs) are receptor tyrosine kinases that upon binding to collagens undergo receptor phosphorylation, which in turn activates signal transduction pathways that regulate cell-collagen interactions. We report here that collagen-dependent DDR1 activation is partly regulated by the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service