HG00BX14
Egg Yolk Emulsion
sterile, emulsion, egg yolk(containing > 50%)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116105
Recommended Products
biological source
egg yolk (containing > 50%)
sterility
sterile
form
emulsion
storage temp.
2-8°C
Application
Used in conjunction with Baird Parker Agar (HG000C41.500) and Potassium Tellurite (HG00BX15.PK)
Preparation Note
Allow to stand at room temperature for 30 minutes prior to use.Shake well
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service