Skip to Content
Merck
All Photos(1)

Key Documents

EX0185

Supelco

Diethyl ether

≥98.0% ethanol basis (diethyl ether), Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Diethyl ether, Diethyl Ether, Ether, Diethyl Ether, Et2O, Ethoxyethane, Ether, Ethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)2O
CAS Number:
Molecular Weight:
74.12
Beilstein:
1696894
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-467-2
grade:
reagent
bp:
34.6 °C (lit.)
vapor pressure:
587 hPa ( 20 °C)

Product Name

Ethyl Ether, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

grade

reagent

Quality Level

Agency

meets USP/NF

vapor density

2.6 (vs air)

vapor pressure

587 hPa ( 20 °C)

Assay

≥98.0% ethanol basis (diethyl ether)

form

liquid

autoignition temp.

320 °F

shelf life

24 mo. (from manufacture date)

expl. lim.

36.5 %

impurities

≤0.0002 meq/g Titrable acid
≤0.001% Carbonyl (as HCHO)
≤0.5% Water (H2O)
≤1 ppm Peroxide (as H2O2)
1-3% Alcohol (C2H5OH)

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n20/D 1.3530 (lit.)

bp

34.6 °C (lit.)

mp

−116 °C (lit.)

transition temp

flash point -40 °C

density

0.706 g/mL at 25 °C (lit.)

shipped in

ambient

storage temp.

2-25°C

SMILES string

CCOCC

InChI

1S/C4H10O/c1-3-5-4-2/h3-4H2,1-2H3

InChI key

RTZKZFJDLAIYFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Expiration dateExp 24 months from mfg dateAlcohol (C2H5OH)1-3%Assay (Et2O + EtOH)98.0% minCarbonyl (as HCHO)0.001% maxColor (APHA)10 maxDensity at 20°C0.714-0.718g/mlEvaporation residue0.001% maxFormClear liquidPeroxide (as H2O2)*1ppm maxTitrable acid0.0002meq/g maxWater (H2O)0.5% max

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service