Skip to Content
Merck
All Photos(1)

Key Documents

EF5013

Sigma-Aldrich

EF5 Antibody Dilution Buffer with Tween 20

EF5 antibody dilution buffer specifically formulated for use with the EF5 Hypoxia Detection Kits and anti-EF5 monoclonal antibodies.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32160801
NACRES:
NA.84

Quality Level

form

liquid

General description

EF5 Antibody Dilution Buffer with Tween 20 is a convenient preparation for the dilution of the anti-EF5 (clone ELK3-51) conjugated antibodies.

Formulated for use with the EF5 Hypoxia Detection Kits:
• EEF5 Hypoxia Detection Kit, Alexa Fluor 488 (Cat. No. EF5-30A4)
• EEF5 Hypoxia Detection Kit, Cyanine 3 (Cat. No. EF5-30C3)

Application

Anti-EF5 antibodies are typically provided at 2 mg/mL. For optimal IHC tissue staining, anti-EF5 antibodies should be diluted with EF5 Antibody Dilution Buffer with Tween 20 to at least 75 ug/mL.
EF5 antibody dilution buffer specifically formulated for use with the EF5 Hypoxia Detection Kits and anti-EF5 monoclonal antibodies.

Components

EF5 Antibody Dilution Buffer with Tween 20: 1 bottle containing 10 mL buffer. Store at -20°C.

Storage and Stability

Store at -20°C.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service