Skip to Content
Merck
All Photos(2)

Key Documents

EC71

Millipore

NovaSeptum® GO Conical Tube Sampling Unit

sterile; γ-irradiated

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104105

material

316L stainless steel cannula
polyester body
polypropylene tube (Fluid contact layer)
polystyrene device
silicone septum (Platinum-cured)
silicone tube (Fluid contact layer: platinum-cured silicone)
silicone tube (medical silicone fluid)
silicone tubing

Quality Level

Agency

according to ISO (sterilization)
meets ISO 14644-1
meets USP 88 Biological Reactivity (all component materials)

sterility

sterile; γ-irradiated

product line

NovaSeptum® GO

feature

autoclavable: no

parameter

0.50 bar max. pressure (7.25 psi)
2-60 °C temp. range (36-140 °F)

fitting

female Luer-Lok® outlet fitting
male Luer-Lok® outlet fitting

application(s)

cell therapy
gene therapy
life science and biopharma
mAb
ophthalmics
parenterals
pharma/biopharma processes
sterile sampling
viral therapy

storage temp.

room temp

General description

The NovaSeptum® GO conical tube sampling unit is intended for applications where high purity, such as low endotoxin levels, is required. The conical tube has a fluid contact layer made of polystyrene making it ideal for endotoxin testing. This sampling unit is available with a 2 mm needle and must be used with a NovaSeptum®GO holder.

This is a single sampling device.

Application

  • Single Sampling
  • Sterile Sampling

Features and Benefits

  • Fluid contact layer made of polystyrene, ideal for endotoxin testing
  • Small size, no need to aliquot

Packaging

EC71-80015: 40 units per box

Components

  • Septum: Platinum-cured silicone
  • Body: Polyester
  • Cannula: ASTM® 316 L Stainless steel
  • Fluid contact layer (tube): Polystyrene, platinum-cured silicone, medical silicone fluid
  • Tubing: Silicone
  • Outlet Tubing: Female and male Luer-Lok®

Other Notes

Assembled under ISO Clean Room Class 8 conditions in a facility certified to ISO 14644-1 standards.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
Luer-Lok is a registered trademark of Becton-Dickinson & Co.
NOVASEPTUM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service