Skip to Content
Merck
All Photos(1)

Key Documents

CP35

Sigma-Aldrich

Anti-PSD-95 (Ab-2) Mouse mAb (7E3-1B8)

liquid, clone 7E3-1B8, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

7E3-1B8, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

rat

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG2a

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

rat ... Dlg4(29495)

General description

Purified mouse monoclonal antibody generated by immunizing BALB/c mice with the specified immunogen and fusing splenocytes with HL-1 mouse myeloma cells (see application references). Recognizes the ~95 kDa PSD-95 protein.
Recognizes the ~95 kDa PSD95 protein.
This Anti-PSD-95 (Ab-2) Mouse mAb (7E3-1B8) is validated for use in Immunoblotting, Immunofluorescence, Immunoprecipitation for the detection of PSD-95 (Ab-2).

Immunogen

Rat
recombinant, rat PSD95

Application


Immunoblotting (1:2000)
Immunofluorescence (1:200, indirect)
Immunoprecipitation (see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Brakeman, P.R., et al. 1997. Nature386, 284.
Hualing, D., et al. 1997. Nature386, 279.
Kennedy, M.B. 1997. Trends in Neurosci.20, 264.
Sanes, J. 1997. Curr. Opin. Neurobiol.7, 93.
Cho., K.-O., et al. 1992. Neuron9, 929.
Immunofluorescent staining of rat hippocampal cells shows a staining pattern coincident with NMDA receptor staining at synaptic sites consistent with PSD-95′s proposed role in receptor clustering. This antibody has also been reported to work for immunoprecipitation. Fixation with cold methanol is recommended for immunofluorescence. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sabrina Reinehr et al.
International journal of molecular sciences, 20(10) (2019-05-30)
Studies have suggested an involvement of the immune system in glaucoma. Hence, a rat experimental autoimmune glaucoma model (EAG) was developed to investigate the role of the immune response. Here, we transferred this model into mice. Either 0.8 mg/mL of
Hanyu Fu et al.
Acta physiologica (Oxford, England), 239(1), e14009-e14009 (2023-06-18)
General anesthesia can induce cognitive deficits in both humans and rodents, correlating with pathological alterations in the hippocampus. However, whether general anesthesia affects olfactory behaviors remains controversial as clinical studies have produced inconsistent results. Therefore, we aimed to investigate how
Jessica L Cifelli et al.
The Journal of biological chemistry, 291(23), 11981-11992 (2016-03-30)
The majority of excitatory synapses in the brain exist on dendritic spines. Accordingly, the regulation of dendritic spine density in the hippocampus is thought to play a central role in learning and memory. The development of novel methods to control
Inmaculada M González-González et al.
Cell reports, 42(5), 112477-112477 (2023-05-07)
Signaling via N-methyl-d-aspartate receptors (NMDARs) is critical for the maturation of glutamatergic synapses, partly through a developmental switch from immature synapses expressing primarily GluN2B- and GluN3A-containing subtypes to GluN2A-rich mature ones. This subunit switch is thought to underlie the synaptic
Han-Yu Fu et al.
Brain pathology (Zurich, Switzerland), 33(2), e13114-e13114 (2022-09-06)
Reactive astrogliosis and neuronal death are major features of brain tissue damage after transient global cerebral ischemia/reperfusion (I/R). The CA1 subfield in the hippocampus is particularly susceptible to cell death after I/R. Recently, attention has focused on the relationship between

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service