CBL212
Anti-Neurofilament 200 kDa Antibody, clone RT97
clone RT97, Chemicon®, from mouse
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
purified antibody
clone
RT97, monoclonal
species reactivity
rat, human, pig, avian, chicken, reptile
species reactivity (predicted by homology)
mammals
manufacturer/tradename
Chemicon®
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
isotype
IgG1
NCBI accession no.
UniProt accession no.
shipped in
wet ice
Specificity
The clone RT97 recognizes the 200 kDa neurofilament polypeptide in a number of mammalian and avian species. Tumours specifically recognized include phaeochromocytoma, paraganglioma, ganglioneuroblastoma and other tumours of neuronal origin.
KNOWN SPECIES CROSS REACTIVITY: Recognizes rat, chicken, pig and reptile
KNOWN SPECIES CROSS REACTIVITY: Recognizes rat, chicken, pig and reptile
Application
Anti-Neurofilament 200 kDa Antibody, clone RT97 detects level of Neurofilament 200 kDa & has been published & validated for use in IH, IH(P).
Research Category
Neuroscience
Neuroscience
Research Sub Category
Neurofilament & Neuron Metabolism
Neuronal & Glial Markers
Neurofilament & Neuron Metabolism
Neuronal & Glial Markers
Staining of both frozen and paraffin wax embedded tissue sections
Strong staining of neuronal processes in a variety of tissues
Optimal working dilutions must be determined by the end user.
Strong staining of neuronal processes in a variety of tissues
Optimal working dilutions must be determined by the end user.
Physical form
Format: Purified
The monoclonal is presented in phosphate buffered saline containing 10mM sodium azide and 1mg/ml bovine serum albumin. We recommend that each laboratory determine an optimum working titre for use in its particular application.
Storage and Stability
Stable for 1 year at 2-8°C from date of receipt
Other Notes
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
recommended
Product No.
Description
Pricing
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Molecular pain, 5, 66-66 (2009-11-20)
Oxaliplatin and related chemotherapeutic drugs cause painful chronic peripheral neuropathies in cancer patients. We investigated changes in neuronal size profiles and neurofilament immunoreactivity in L5 dorsal root ganglion (DRG) tissue of adult female Wistar rats after multiple-dose treatment with oxaliplatin
Signals for the initiation and termination of synthesis of the viral strand of bacteriophage f1.
Cold Spring Harbor Symposia on Quantitative Biology, 47 Pt 2, 717-722 (1983)
2-(Cyclohexylamino)-1-(4-cyclopentylpiperazin-1-yl)-2-methylpropan-1-one, a novel compound with neuroprotective and neurotrophic effects in vitro.
Neurochemistry International null
Tissue engineering a complete vaginal replacement from a small biopsy of autologous tissue.
Transplantation null
Neuro-degenerative diseases, 17(4-5), 181-198 (2017-05-11)
A novel ataxic mouse line was established from the offspring of a male mouse administered cyclophosphamide in a juvenile period. We have attempted to examine the phenotype and histopathological changes of affected mice. Furthermore, linkage analysis and sequencing of the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service