Skip to Content
Merck
All Photos(2)

Key Documents

CBL171

Sigma-Aldrich

Anti-α-Smooth Muscle Actin (ACTA2) Antibody,

CHEMICON®, mouse monoclonal, ASM-1

Synonym(s):

Alpha-actin-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Actin Antibody, smooth muscle, clone ASM-1, clone ASM-1, Chemicon®, from mouse

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

ASM-1, monoclonal

species reactivity

chicken, mouse, horse, rat, human, bovine

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ACTA2(59)

General description

ASM-1 represents an excellent marker for myogenic soft tissue tumors and smooth muscle differentiation. This antibody reacts with many types of smooth muscle cells, such as those present in vascular walls, intestinal muscularis mucosae and propria, myometrium, stroma of various tissues, and is also positive for myoepithelial cells of various glands, notably salivary and mammary gland. Myogenic soft tissues detected include leiomyosarcomas, leiomyomas, and certain stromal cells surrounding infiltrating ductal carcinoma of the breast.

Specificity

Expected to cross-react with bovine, equine and chicken. Reactivity with other species has not been confirmed.
This antibody is specific for the alpha-smooth-muscle isoform of actin (MW 43 kDa).

Immunogen

Epitope: N-terminus
Synthetic peptide corresponding to the ten N-terminal amino acids of the alpha-smooth muscle isoform of actin.

Application

Anti-Actin Antibody, smooth muscle, clone ASM-1 detects level of Actin & has been published & validated for use in IF, IH, IH(P) & WB.
Immunohistochemistry:
1:2000 dilution of a previous lot was used on frozen and formalin-fixed, paraffin-embedded tissues; protease pretreatment is recommended for paraffin-embedded sections.

Immunofluorescence:
A previous lot of this antibody was used on Immunofluoresence.

Optimal working dilutions must be determined by the end user.
Research Category
Cell Structure
Research Sub Category
Cytoskeletal Signaling

Quality

Evaluated by Western Blot on HUVEC lysates.

Western Blot Analysis:
1:1000 dilution of this antibody detected ACTIN, SM MUSC on 10 μg of HUVEC lysates.

Target description

43 kDa

Linkage

Replaces: 04-1040

Physical form

Format: Purified
Protein A purified
Purified mouse monoclonal IgG2a presented as lyophilized material. Reconstitute in 1 mL of of distilled water.
Final solution is PBS, pH 7.4, containing 0.5% BSA and 0.09% sodium azide.

Storage and Stability

Stable for 1 year at -20°C in undiluted aliquots from date of receipt.
Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Positive reaction using stress fibers of smooth muscle-derived cells and some smooth muscle subtype fibroblasts.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reprogramming LCLs to iPSCs Results in Recovery of Donor-Specific Gene Expression Signature.
Thomas, SM; Kagan, C; Pavlovic, BJ; Burnett, J; Patterson, K; Pritchard, JK; Gilad, Y
PLoS Genetics null
Manuela Mura et al.
Stem cell research, 39, 101510-101510 (2019-08-10)
We generated PSMi001-A and PSMi008-A hiPSC lines from two individuals belonging to a South African (SA) founder population in which the malignant KCNQ1-A341V mutation cosegregates with the Long QT Syndrome (LQTS) phenotype. PSMi001-A was derived from an asymptomatic KCNQ1-A341V mutation
Localization of angiopoietin-1 and Tie2 immunoreactivity in rodent ependyma and adjacent blood vessels suggests functional relationships.
Horton, BN; Solanki, RB; Rajneesh, KF; Kulesza, P; Ardelt, AA
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society null
Constantin C Chipev et al.
BMC dermatology, 2, 13-13 (2002-11-26)
Wounds in the nonglabrous skin of keloid-prone individuals tend to cause large disordered accumulations of collagen which extend beyond the original margins of the wound. In addition to abnormalities in keloid fibroblasts, comparison of dermal fibroblasts derived from nonwounded glabrous
Zhaocheng Zhang et al.
Cell death & disease, 12(7), 644-644 (2021-06-26)
Dental pulp stem cells (DPSC) are capable of differentiating into vascular endothelial cells. Although the capacity of vascular endothelial growth factor (VEGF) to induce endothelial differentiation of stem cells is well established, mechanisms that maintain stemness and prevent vasculogenic differentiation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service