Skip to Content
Merck
All Photos(1)

Key Documents

CBL101

Sigma-Aldrich

Mouse Anti-Human IgG Antibody, Fc, all subclasses, clone MK1A6

clone MK1A6, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

mouse

Quality Level

100
300

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

MK1A6, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
inhibition assay: suitable (hemagglutination)

isotype

IgG1

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

Specificity

This antibody is specific for the CH2 domain of all subclasses of the Fc region of human IgG

FUSION PARTNER: NSO myeloma cell line

Immunogen

Polyclonal human IgG

Application

ELISA assay development of total IgG

Passive haemagglutination assays

Histological staining of acetone fixed frozen tissue sections

Optimal working dilutions must be determined by the end user.
Mouse anti-Human IgG Antibody, Fc, all subclasses, clone MK1A6 detects level of Human IgG & has been published & validated for use in ELISA, IF, HI, IH.
Research Category
Secondary & Control Antibodies
Research Sub Category
Fragment Specific Secondary Antibodies

Linkage

Replaces: MABN1055

Physical form

Format: Purified
The monoclonal was purified by protein A affinity chromatography and is presented as a liquid in phosphate buffered saline containing 10mM sodium azide.

Storage and Stability

For use within 1 month of purchase store at +4°C, for long term storage aliquot antibody into small volumes and store at -20°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David R McIlwain et al.
Cell host & microbe, 29(12), 1828-1837 (2021-11-17)
Developing new influenza vaccines with improved performance and easier administration routes hinges on defining correlates of protection. Vaccine-elicited cellular correlates of protection for influenza in humans have not yet been demonstrated. A phase-2 double-blind randomized placebo and active (inactivated influenza

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service