Skip to Content
Merck
All Photos(1)

Key Documents

AP192P

Sigma-Aldrich

Donkey Anti-Mouse IgG Antibody, HRP conjugate, Species Adsorbed

Chemicon®, from donkey

Synonym(s):

Anti-Mouse antibody, Donkey anti-Mouse IgG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

donkey

Quality Level

conjugate

peroxidase conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

Specificity

Specific for mouse IgG, heavy and light chain.

Application

Donkey anti-Mouse IgG Antibody, HRP conjugate, Species Adsorbed is an antibody against Donkey Mouse IgG for use in ELISA & WB.
EIA and Western blots: 1:5,000-1:100,000.

Immunohistochemistry: 1:500-1:5,000.

Optimal working dilutions must be determined by end user.
Research Category
Secondary & Control Antibodies
Research Sub Category
Secondary Antibodies Adsorbed for Dual Labeling

Physical form

Immunoaffinity Purified and Immunodepletion
Lyophilized. Buffer = 0.01 M Sodium Phosphate, 0.15M NaCl, pH 7.1 with 15 mg/mL BSA.

RECONSTITUTION:

Reconstitute with 500 μL of sterile distilled water.

Storage and Stability

Maintain lyophilized product at 2–8°C for up to 12 months. After reconstitution the product is stable for several weeks at 2–8°C as an undiluted liquid. For extended storage after reconstitution, add an equal volume of glycerol to make a final concentration of 50% glycerol followed by storage at -20°C in undiluted aliquots for up to 12 months. Please note the concentration of protein (and buffer salts) will decrease to one-half of the original after the addition of glycerol. Avoid repeated freeze/thaw cycles.

WARNING:

Use of sodium azide as a preservative will substantially inhibit the enzyme activity of HRP.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dongmei Hu et al.
Biosensors, 3(3), 297-311 (2013-01-01)
Two label-free biosensor platforms, Resonance Waveguide Grating (RWG) and Surface Plasmon Resonance (SPR), were used to rank a large panel of anti-dengue virus NS1 antibodies. Dengue non-structural 1 (NS1) protein is an established serological marker for the early detection of
Hae-Young Park et al.
NPJ vaccines, 2, 31-31 (2017-12-22)
Targeting model antigens (Ags) to Clec9A on DC has been shown to induce, not only cytotoxic T cells, but also high levels of Ab. In fact, Ab responses against immunogenic Ag were effectively generated even in the absence of DC-activating
Decorin induces rapid secretion of thrombospondin-1 in basal breast carcinoma cells via inhibition of Ras homolog gene family, member A/Rho-associated coiled-coil containing protein kinase 1.
Neill, T; Jones, HR; Crane-Smith, Z; Owens, RT; Schaefer, L; Iozzo, RV
FEBS Journal null
Hiroyuki Honda et al.
Journal of neuropathology and experimental neurology, 75(11), 1008-1019 (2016-09-17)
Prion protein (PrP) has 2 glycosylated sites and a glycosylphosphatidylinositol (GPI) anchor on the C-terminal. Reports on genetic prion disease with GPI anchorless PrP are very limited. In this study, we characterized the molecular alterations of mutated PrP in a
Abrey J Yeo et al.
Cell discovery, 1, 15025-15025 (2015-01-01)
Senataxin, defective in ataxia oculomotor apraxia type 2, protects the genome by facilitating the resolution of RNA-DNA hybrids (R-loops) and other aspects of RNA processing. Disruption of this gene in mice causes failure of meiotic recombination and defective meiotic sex

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service