Skip to Content
Merck
All Photos(1)

Key Documents

AP175

Sigma-Aldrich

Goat Anti-Human IgE Antibody

3.1 mg/mL, Chemicon®

Synonym(s):

Anti-Human IgE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

concentration

3.1 mg/mL

technique(s)

ELISA: suitable
immunoprecipitation (IP): suitable
western blot: suitable

shipped in

dry ice

target post-translational modification

unmodified

Specificity

Human IgE. Single arc by IEP against IgE elevated serum. Less than 1% reactivity with IgM, IgG, IgD and IgA.

Immunogen

Human myeloma IgE

Application

Goat anti-Human IgE Antibody is an antibody against Human IgE for use in ELISA, IP & WB.

Physical form

Immunoglobulin, affinity purified. Liquid in 0.05M Tris-HCl (pH 7.5), 0.5M NaCl, 0.01M EDTA with 0.1% sodium azide.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Blankestijn et al.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology, 48(9), 1206-1213 (2018-06-16)
The role of sensitization to commercially available allergens of English walnut (Juglans regia) Jug r 1, 2 and 3 in walnut allergy has been previously investigated in walnut allergic adults and was unable to explain all cases of walnut allergy.
Primary respiratory and food allergy to mealworm.
Henrike C H P Broekman et al.
The Journal of allergy and clinical immunology, 140(2), 600-603 (2017-03-10)
A designer cell-based histamine-specific human allergy profiler.
Auslander, D; Eggerschwiler, B; Kemmer, C; Geering, B; Auslander, S; Fussenegger, M
Nature Communications null
Elin Rönnberg et al.
International archives of allergy and immunology, 165(1), 68-74 (2014-10-25)
Many of the functions attributed to mast cells depend on the various pro-inflammatory mediators that are secreted upon mast cell activation. These include a panel of mast cell-specific proteases. In addition, recent studies have indicated that murine mast cells also
Mervi Alanne-Kinnunen et al.
PloS one, 9(9), e108352-e108352 (2014-09-25)
Activated mast cells in atherosclerotic lesions degranulate and release bioactive compounds capable of regulating atherogenesis. Here we examined the ability of activated human primary mast cells to regulate the expression of the major scavenger receptors in cultured human primary monocyte-derived

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service