Skip to Content
Merck
All Photos(1)

Key Documents

AP108B

Sigma-Aldrich

Goat Anti-Guinea Pig IgG Antibody, biotin conjugate

Chemicon®, from goat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

biotin conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

guinea pig

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Application

Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies
This Goat anti-Guinea Pig IgG Antibody, biotin conjugate is validated for use in ELISA, WB for the detection of Guinea Pig IgG.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diyi Zhou et al.
Molecular medicine reports, 23(1) (2020-11-26)
Pancreatic and duodenal homeobox (PDX)‑1 is a gene that plays an important role in pancreatic development and function. Type‑2 diabetes mellitus (T2DM) is a metabolic disease associated with insulin resistance and impaired islet β‑cell function. There is evidence that methylation
Jie Qi et al.
Frontiers in molecular neuroscience, 11, 281-281 (2018-08-31)
Sulbactam is an atypical β-lactam medication and reported to be neuroprotective by up-regulating glial glutamate transporter-1 (GLT-1) in rats. The present study was undertaken to study the role of p38 MAPK signal pathway in sulbactam induced up-regulation of GLT-1 expression
Eriko Kuramoto et al.
The Journal of comparative neurology, 525(18), 3821-3839 (2017-09-02)
The rodent orbitofrontal cortex is involved in a variety of cognitive and behavioral functions that require thalamic input to be successfully expressed. Although the thalamic nucleus submedius (Sm) is a major source of afferents to the orbitofrontal cortex, thalamocortical projection

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service