Skip to Content
Merck
All Photos(7)

Key Documents

ABT1487

Sigma-Aldrich

Anti-alpha-Actin Antibody, Smooth Muscle

from rabbit, purified by affinity chromatography

Synonym(s):

Actin, aortic smooth muscle, Alpha-actin-2, Cell growth-inhibiting gene 46 protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse, human

species reactivity (predicted by homology)

chicken (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ACTA1(58)

General description

Actin, aortic smooth muscle (UniProt P62736; also known as Alpha-actin-2, Cell growth-inhibiting gene 46 protein) is encoded by the ACTA2 (also known as AAT6, ACTSA, ACTVS, GIG46, MYMY5) gene (Gene ID 59) in human. Actins are globular multi-functional proteins that serve as the basic building blocks of cytoskeletal microfilaments and are among the most conserved eukaryotic proteins. Six actin types exist, skeletal muscle alpha-actin is encoded by the ACTA1 gene, smooth muscle alpha-actin by the ACTA2 gene, cytoplasmic beta-actin by the ACTB gene, cardiac muscle alpha-actin by the ACTC1 gene, cytoplasmic gamma-actin by the ACTG1 gene, and smooth muscle gamma-actin by the ACTG2 (a.k.a. ACTA3) gene. Although actins show >90% overall sequence homology, isoforms do show spatial, temporal, and tissue-specific expresson patterns and only 50-60% homology is found in their 18 N-terminal residues. Cytoplasmic β and γ-actins, are thought to be present in all cells, while the other four actin isoforms are typically found in specific adult muscle tissue types. Actins exist in a variety of structural states, depending on the specific ionic conditions or the interaction with ligand proteins. The oligomeric and polymeric forms that actin molecules assume are dependent on the distinct conformations they adopt.

Specificity

This polyclonal antibody targets the N-terminal sequence specific to smooth muscle alpha-actin (ACTA2), but not skeletal muscle alpha-actin (ACTA1), cytoplasmic beta-actin (ACTB), cardiac muscle alpha-actin (ACTC1), cytoplasmic gamma-actin (ACTG1), or smooth muscle gamma-actin (ACTG2; ACTA3).

Immunogen

Epitope: N-terminus.
KLH-conjugated linear peptide corresponding to the N-terminual sequence of human smooth muscle alpha-actin.

Application

Anti-alpha-Actin Antibody, Smooth Muscle is an antibody against alpha-Actin for use in Western Blotting, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation.
Immunocytochemistry Analysis: 5.0 µg/mL from a representative lot immunostained smooth muscle alpha-Actin in NIH/3T3 fibroblasts, HUVECs, A431 and HeLa cells.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot immunostained smooth muscle cells in human uterus tissue sections.
Immunoprecipitation Analysis: 10 µg from a representative lot immunoprecipitated smooth muscle alpha-Actin from 500 µg of C2C12 mouse myoblast lysate.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton

Quality

Evaluated by Western Blotting in C2C12 mouse myoblast lysate.

Western Blotting Analysis: 0.25 µg/mL of this antibody detected smooth muscle alpha-Actin in 10 µg of C2C12 mouse myoblast lysate.

Target description

~42 kDa observed.

Physical form

Affinity purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zaki Hakami et al.
Anatomical record (Hoboken, N.J. : 2007), 301(11), 1820-1833 (2018-10-06)
Although minor salivary glands play a significant functional role in the oral cavity, their developmental morphology and cell differentiation has been scarcely studied. This study aimed to describe the development of rat palatine glands with regard to the ultrastructural morphology
Christopher L Robinson et al.
Nature communications, 9(1), 2090-2090 (2018-05-31)
Evidence of male-to-female sexual transmission of Zika virus (ZIKV) and viral RNA in semen and sperm months after infection supports a potential role for testicular cells in ZIKV propagation. Here, we demonstrate that germ cells (GCs) are most susceptible to
Virender Kumar et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 26(12), 2798-2811 (2018-10-06)
Trans-differentiation of quiescent hepatic stellate cells (HSCs) into active myofibroblasts secretes excess amounts of extracellular matrix (ECM) proteins. miR-29b1 has the potential to treat liver fibrosis, because it targets several profibrotic genes. We previously demonstrated that miR-29b1 and the hedgehog
Jian Chen et al.
Scientific reports, 6, 30217-30217 (2016-07-28)
Disruption of the TGF-β pathway is associated with liver fibrosis and suppression of liver tumorigenesis, conditions associated with low Vitamin D (VD) levels. However, potential contributions of VD to liver tumor progression in the context of TGF-β signaling remain unexplored.
David Carr et al.
Breast cancer research : BCR, 24(1), 40-40 (2022-06-11)
The Bcl-3 protein is an atypical member of the inhibitor of -κB family that has dual roles as a transcriptional repressor and a coactivator for dimers of NF-κB p50 and p52. Bcl-3 is expressed in mammary adenocarcinomas and can promote

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service