Skip to Content
Merck
All Photos(3)

Key Documents

ABT1485

Sigma-Aldrich

Anti-Actin

from rabbit, purified by affinity chromatography

Synonym(s):

Actin, alpha skeletal muscle, Alpha-actin-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... ACTA1(58)

General description

Actins are globular multi-functional proteins that serve as the basic building blocks of cytoskeletal microfilaments and are among the most conserved eukaryotic proteins. Six actin types exist, skeletal muscle alpha-actin (ACTA1), smooth muscle alpha-actin (ACTA2), cytoplasmic beta-actin (ACTB), cardiac muscle alpha-actin (ACTC1), cytoplasmic gamma-actin (ACTG1), and smooth muscle gamma-actin (ACTG2, a.k.a. ACTA3). Although actins show >90% overall sequence homology, isotypes do show spatial, temporal, and tissue-specific expresson patterns and only 50-60% homology is found in their 18 N-terminal residues. Cytoplasmic - and -actins are thought to be present in all cells, while the other four actin isoforms are typically found in specific adult muscle tissue types. Actins exist in a variety of structural states, depending on the specific ionic conditions or the interaction with ligand proteins. The oligomeric and polymeric forms that actin molecules assume are dependent on the distinct conformations they adopt. In additon to serving as the basic building blocks of cytoskeletal microfilaments, actins are involved in multiple nuclear functions and adopt specific conformations that are important for their association with nuclear actin-binding proteins.

Specificity

Broad species reactivity expected due to 100% sequence homology, including Bovine, Horse, Non-Human Primate, Porcine, Rabbit, and Rat.
This rabbit polyclonal antibody was raised against skeletal muscle alpha-actin (ACTA1) N-terminal end sequence that shares 80% sequence identity in the corresponding region with cardiac muscle alpha-actin (ACTC1), smooth muscle alpha-actin (ACTA2), and cytoplasmic gamma-actin (ACTG1). Immunogen sequence is not found in smooth muscle gamma-actin (ACTG2, a.k.a. ACTA3) and cytoplasmic beta-actin (ACTB).

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to the N-terminal end sequence of mature skeletal muscle alpha-actin (ACTA1).

Application

Immunocytochemistry Analysis: A 1:250 dilution from a representative lot detected both cytoplasmic and nuclear actin immunoreactivity in A431 and NIH/3T3 cells.

Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected actin in human cardiac muscle, skeletal muscle, and small intestine tissue sections.

This polyclonal antibody is also available as Alexa Fluor 488 and Alexa Fluor 647 conjugates (Cat. No. ABT1485-AF488 and ABT1485-AF647) for flow cytometry, immunocytochemistry, immunofluorescence applications.
Research Category
Cell Structure
This rabbit polyclonal Anti-Actin Antibody, Cat. No. ABT1485 is validated for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting for the detection of Actin.

Quality

Evaluated by Western Blotting in human skeletal muscle tissue lysate.

Western Blotting Analysis: A 1:5,000 dilution of this antibody detected actin in 10 µg of human skeletal muscle tissue lysate.

Target description

~42.0 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity purified.
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuzuha Shiraki et al.
Molecular biology of the cell, 34(11), ar110-ar110 (2023-08-16)
Alcadein α (Alcα) and amyloid-β protein precursor (APP) are cargo receptors that associate vesicles with kinesin-1. These vesicles, which contain either Alcα or APP, transport various proteins/cargo molecules into axon nerve terminals. Here, we analyzed immune-isolated Alcα- and APP-containing vesicles
Rosanna Maccari et al.
European journal of medicinal chemistry, 252, 115270-115270 (2023-03-20)
Type 2 diabetes mellitus (T2DM) is a serious chronic disease with an alarmingly growing worldwide prevalence. Current treatment of T2DM mainly relies on drug combinations in order to control blood glucose levels and consequently prevent the onset of hyperglycaemia-related complications.
Rosaria Ottanà et al.
Molecules (Basel, Switzerland), 26(2) (2021-01-14)
Diabetes mellitus (DM) is a complex disease which currently affects more than 460 million people and is one of the leading cause of death worldwide. Its development implies numerous metabolic dysfunctions and the onset of hyperglycaemia-induced chronic complications. Multiple ligands
Yaofeng Zhu et al.
International journal of molecular medicine, 44(4), 1414-1424 (2019-08-01)
The balance between glutamate (cortex and thalamus) and dopamine (substantia nigra) inputs on striatal neurons is of vital importance. Dopamine deficiency, which breaks this balance and leads to the domination of cortical glutamatergic inputs, plays an important role in Parkinson's

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service