Skip to Content
Merck
All Photos(3)

Key Documents

ABT135

Sigma-Aldrich

Anti-phospho-FAK (Tyr397) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Focal adhesion kinase 1, FADK 1, Protein-tyrosine kinase 2, pp125FAK

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

species reactivity (predicted by homology)

human (based on 100% sequence homology), mouse (based on 100% sequence homology), Xenopus (based on 100% sequence homology), chicken (based on 100% sequence homology)

technique(s)

cell based assay: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr397)

Gene Information

human ... PTK2(5747)

General description

Focal adhesion kinase (FAK) is a non receptor protein tyrosine kinase discovered as a substrate for Src and a key element in integrin signaling. FAK plays a central role in cell spreading, differentiation, migration, cell death and acceleration of the G1 to S phase transition of the cell cycle. FAK regulation includes phosphorylation at multiple tyrosine and serine residues. Phosphorylation of tyrosine is generally associated with positive regulation and growth promotion; however, dephosphorylation at these sites occurs as cells enter mitosis (M-Phase). In contrast, serine phosphorylation either remains high or is increased as cells enter mitosis and may play a role in focal adhesion disassembly. FAK and its phosphorylation states have been implicated in cancer metastasis, tumor cell survival, and adhesion-independent growth. Recent evidence indicates that elevation of FAK activity in human carcinoma cells is associated with increased invasive potential. A central role in tumor formation and progression suggests that FAK is an attractive target for therapeutic intervention. Activation of FAK by integrin clustering leads to autophosphorylation at Tyr397.

Specificity

This antibody recognizes FAK phosphorylated at Tyr397.

Immunogen

Epitope: Phosphorylated Tyr397
KLH-conjugated linear peptide corresponding to human FAK phosphorylated at Tyr397.

Application

Detect phospho-FAK (Tyr397) using this Anti-phospho-FAK (Tyr397) Antibody validated for use in WB, Cell Function Assay.
Peptide Inhibition Assay Analysis: A 1:1,000 dilution from a representative lot peptide blocked on RAT2 cells transfected with Src.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)

Quality

Evaluated by Western Blot in untransfected and Src-transfected RAT2 cell lysates.

Western Blot Analysis: A 1:1,000 dilution of this antibody detected phospho-FAK (Tyr397) in 10 µg of untransfected and Src-transfected RAT2 cell lysates.

Target description

~125 kDa observed.
An uncharacterized band appears at ~190 kDa in some lysates.

Linkage

Replaces: 04-974

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
untransfected and Src-transfected RAT2 cell lysates

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer J Chia et al.
The Journal of clinical investigation, 126(11), 4331-4345 (2016-11-02)
Scleroderma is a group of skin-fibrosing diseases for which there are no effective treatments. A feature of the skin fibrosis typical of scleroderma is atrophy of the dermal white adipose tissue (DWAT). Adipose tissue contains adipose-derived mesenchymal stromal cells (ADSCs)
Won-Sik Shin et al.
Journal of cellular biochemistry, 118(9), 2887-2895 (2017-02-19)
Protein tyrosine kinase 6 (PTK6; also known as Brk) is closely related to the Src family kinases, but lacks a membrane-targeting myristoylation signal. Sublocalization of PTK6 at the plasma membrane enhances its oncogenic potential. To understand the mechanism(s) underlying the
Jae Hoon Kim et al.
International journal of molecular sciences, 23(20) (2022-10-28)
PTK7 is a catalytically defective receptor protein tyrosine kinase upregulated in various cancers, including esophageal squamous cell carcinoma (ESCC). In previous studies, we observed a positive correlation between PTK7 expression levels and tumorigenicity in various ESCC cell lines and xenograft
Won-Sik Shin et al.
International journal of molecular sciences, 23(4) (2022-02-27)
Protein tyrosine kinase 7 (PTK7), a catalytically defective receptor protein tyrosine kinase, is upregulated in tumor tissues and cell lines of esophageal squamous cell carcinoma (ESCC). We showed that PTK7 plays an oncogenic role in various ESCC cell lines. However
Hua-Li Yu et al.
International journal of molecular sciences, 25(13) (2024-07-13)
Microglia migrate to the cerebral cortex during early embryonic stages. However, the precise mechanisms underlying microglia migration remain incompletely understood. As an extracellular matrix protein, Netrin-1 is involved in modulating the motility of diverse cells. In this paper, we found

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service