Skip to Content
Merck
All Photos(4)

Key Documents

ABS233

Sigma-Aldrich

Anti-PI3 Kinase p85 Antibody, N-SH2 domain

from rabbit, purified by affinity chromatography

Synonym(s):

Phosphatidylinositol 3-kinase regulatory subunit alpha, PI3-kinase regulatory subunit alpha, PI3K regulatory subunit alpha, PtdIns-3-kinase regulatory subunit alpha, Phosphatidylinositol 3-kinase 85 kDa regulatory subunit alpha, PI3-kinase subunit p85-al

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), monkey (based on 100% sequence homology), bovine (based on 100% sequence homology)

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

bovine ... Pik3R1(282307)
human ... PIK3R1(5295)
mouse ... Pik3R1(18708)
rat ... Pik3R1(25513)
rhesus monkey ... Pik3R1(698996)

General description

Phosphatidylinositol 3-kinase regulatory subunit alpha (UniProt Q63787; also known as Phosphatidylinositol 3-kinase 85 kDa regulatory subunit alpha, PI3-kinase regulatory subunit alpha, PtdIns-3-kinase regulatory subunit p85-alpha, PI3-kinase subunit p85-alpha, PI3K regulatory subunit alpha, PtdIns-3-kinase regulatory subunit alpha) is encoded by the Pik3r1 gene (Gene ID 25513) in rat species. PI3K regulatory subunit alpha catalyzes the phosphorylation of PI(4,5)P2 inositol ring at the 3 position to generate PI(3,4,5)P3, a potent second messenger that mediates survival signaling and insulin action. PI3 Kinase is a heterodimeric complex composed of an 85 kDa regulatory subunit and a 110 kDa catalytic subunit. Following ligand binding, tyrosine phosphorylation of growth factor receptors creates docking sites for p85 binding (through its SH2 domains), and acts as an adapter, mediating the association of the p110 catalytic unit to the plasma membrane. Plays a role in insulin-stimulated increase in glucose uptake and glycogen synthesis in insulin-sensitive tissues. Mutations in Pik3r1 gene are known to be associated with insulin resistance.

Specificity

This polyclonal antibody targets the first SH2 domain region common to human (isoform 1, AS53, p46, p85I), mouse, and rat (isoform p85-alpha, p55-alpha, and p50-alpha) spliced isoforms of PI3 kinase regulatory subunit alpha reported by UniProt (P27986, P26450, Q63787).

Immunogen

Epitope: First SH2 domain.
GST-tagged recombinant fragment corresponding to the first SH2 domain of rat PI3 kinase regulatory subunit alpha.

Application

Detect PI3 Kinase regulatory subunit alpha using this rabbit polyclonal Anti-PI3 Kinase p85, N-SH2 domain, Cat. No. ABS233, validated for use in Immunoprecipitation and Western Blotting.
Immunoprecipitation Analysis: 5 µg from a representative lot immunoprecipitated PI3 kinase regulatory subunit alpha isoforms from 200 µg of rat brain tissue lysate.

Immunoprecipitation Analysis: 5 µg from a representative lot immunoprecipitated PI3 kinase regulatory subunit alpha isoforms from 500 µg of Jurkat cell lysate.

Western Blotting Analysis: 2 µg/mL from a representative lot detected PI3 kinase regulatory subunit alpha isoforms in 10 µg of Jurkat cell lysate.
Research Category
Signaling
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in rat brain tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected PI3 kinase regulatory subunit alpha isoforms in 10 µg of rat brain tissue lysate.

Target description

~85/50 kDa observed. 83.47/53.49/49.97/84.47 kDa (human isoform 1/AS53/p46/p85I; UniProt P27986), 83.39 kDa (mouse; UniProt P26450), 83.40/53.39/50.00 kDa (rat p85-alpha/p55-alpha/p50-alpha isoforms; UniProt Q63787). Uncharacterized bands may be observed in some lysate(s).

Linkage

Replaces: 06-496

Physical form

Affinity purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Jurkat cell lysate

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu-Jie Hu et al.
Nucleic acids research, 43(16), 7790-7804 (2015-06-29)
Jumonji domain-containing protein 6 (JMJD6) is a nuclear protein involved in histone modification, transcription and RNA processing. Although JMJD6 is crucial for tissue development, the link between its molecular functions and its roles in any given differentiation process is unknown.
Teresita Padilla-Benavides et al.
International journal of molecular sciences, 21(3) (2020-02-06)
Brg1 (Brahma-related gene 1) is one of two mutually exclusive ATPases that can act as the catalytic subunit of mammalian SWI/SNF (mSWI/SfigureNF) chromatin remodeling enzymes that facilitate utilization of the DNA in eukaryotic cells. Brg1 is a phospho-protein, and its
Asif J Iqbal et al.
eLife, 5 (2016-08-31)
Apolipoprotein A1 (apoA1) is the major protein component of high-density lipoprotein (HDL) and has well documented anti-inflammatory properties. To better understand the cellular and molecular basis of the anti-inflammatory actions of apoA1, we explored the effect of acute human apoA1
Jonathon N Winnay et al.
The Journal of clinical investigation, 126(4), 1401-1412 (2016-03-15)
The phosphatidylinositol 3-kinase (PI3K) signaling pathway is central to the action of insulin and many growth factors. Heterozygous mutations in the gene encoding the p85α regulatory subunit of PI3K (PIK3R1) have been identified in patients with SHORT syndrome - a
Yu-Jie Hu et al.
F1000Research, 2, 279-279 (2013-01-01)
Protein arginine methylation is a common posttranslational modification that has been implicated in numerous biological processes including gene expression. The mammalian genome encodes nine protein arginine methyltransferases (Prmts) that catalyze monomethylation, asymmetric dimethylation, and symmetric dimethylation on arginine residues. Protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service