Skip to Content
Merck
All Photos(2)

Key Documents

ABE1007

Sigma-Aldrich

Anti-BATF3 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Basic leucine zipper transcriptional factor ATF-like 3, B-ATF-3, 21 kDa small nuclear factor isolated from T-cells, Jun dimerization protein p21SNFT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

flow cytometry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BATF3(55509)

General description

BATF3, also known as Basic leucine zipper transcriptional factor ATF-like 3, B-ATF-3, 21 kDa small nuclear factor isolated from T-cells, or Jun dimerization protein p21SNFT, and encoded by the gene BATF3/SNFT, is an AP-1 family transcription factor that controls the differentiation of CD8+ thymic conventional dendritic cells in the immune system. BATF3 is required for the development of CD8-alpha+ classical dendritic cells (cDCs) and related CD103+ dendritic cells that cross-present antigens to CD8. BATF3 acts via the formation of a heterodimer with JUN family proteins that recognizes and bind the DNA sequence 5′-TGA[CG]TCA-3′ and thus regulates the expression of the target genes. BATF3 is localized to the nucleus. BATF3 is expressed in myeloid dendritic cells and T cell precursor immune cells primarily.

Immunogen

Epitope: Leucine-zipper
KLH-conjugated linear peptide corresponding to the Leucine-zipper of human BATF3.

Application

Detect BATF3 using this Anti-BATF3 antibody validated for use in western blotting & flow cytometry.
Flow Cytometry Analysis: 1 µg of this antibody detected BATF3 in human PBMC cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors

Quality

Evaluated by Western Blotting in human spleen tissue lysate.

Western Blotting Analysis: 0.25 µg/mL of this antibody detected BATF3 in 10 µg of human spleen tissue lysate.

Target description

~17 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Wang et al.
Molecular immunology, 99, 104-114 (2018-05-11)
Basic leucine zipper transcription factor ATF-like (BATF) -3 is a member of the activator protein 1 (AP‑1) family of transcription factors and is known to play a vital role in regulating differentiation of antigen-presenting cells in mammals. In this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service