Skip to Content
Merck
All Photos(2)

Key Documents

AB9220

Sigma-Aldrich

Anti-Nanog Antibody

serum, Chemicon®

Synonym(s):

Anti-NANOG/STM1, Anti-STM1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
western blot: suitable

input

sample type: mouse embryonic stem cell(s)
sample type induced pluripotent stem cell(s)
sample type: human embryonic stem cell(s)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NANOG(79923)
mouse ... Nanog(71950)

Related Categories

General description

NANOG is a gene expressed in embryonic stem cells (ESCs) and is thought to be a key factor in maintaining pluripotency. NANOG thought to function in concert with other factors such as POU5F1 and SOX2 to establish ESC identity. These cells offer an important area of study because of their ability to maintain pluripotency. In other words, these cells have the ability to become virtually any cell of any of the three germ layers (endoderm, ectoderm, mesoderm).

Specificity

Recognizes Nanog, human.

Immunogen

Synthetic peptide from human Nanog.

Application

Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation
This Anti-Nanog Antibody is validated for use in FC, WB for the detection of Nanog.
Western blotting: 1:1,000-1:5,000. The antibody recognizes a protein of ~35 kDa on MBL-5 cell lysate. Additional bands of ~50 and ~60 kDa may be seen depending on sample and antibody dilution used.
Flow Cytometry: Cormier et al., 2006. Tissue Engineering 12:3233-3245.

Optimal working dilutions must be determined by the end user.

Target description

39 kDa (MW 34 kDa)

Physical form

Rabbit Serum. Liquid.
Unpurified

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Whole cell lysate from mouse embryonic stem cells

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A genome-wide RNAi screen identifies a new transcriptional module required for self-renewal.
Hu, G; Kim, J; Xu, Q; Leng, Y; Orkin, SH; Elledge, SJ
Genes & Development null
Matthew J Simmons et al.
Breast cancer research : BCR, 14(5), R126-R126 (2012-09-21)
NOTCH activation has been recently implicated in human breast cancers, associated with a poor prognosis, and tumor-initiating cells are hypothesized to mediate resistance to treatment and disease relapse. To address the role of NOTCH1 in mammary gland development, transformation, and
Difei Dong et al.
Gene expression, 15(4), 153-162 (2012-07-13)
FoxA1 belongs to the fork head/winged-helix transcription factor family and participates in stimulating neuronal differentiation of pluripotent stem cells at early stages. To explore the biological roles of FoxA1 during this process, the stable expression of a GFP-FoxA1 fusion protein
Miguel Mendivil-Perez et al.
International journal of molecular sciences, 24(10) (2023-05-27)
Alzheimer's disease (AD) is a chronic neurological condition characterized by the severe loss of cholinergic neurons. Currently, the incomplete understanding of the loss of neurons has prevented curative treatments for familial AD (FAD). Therefore, modeling FAD in vitro is essential
Arlette A Haidar-Montes et al.
Animals : an open access journal from MDPI, 14(15) (2024-08-10)
Adipose-derived stem cells (ADSCs) hold promise for tendon repair, even if their tenogenic plasticity and underlying mechanisms remain only partially understood, particularly in cells derived from the ovine animal model. This study aimed to characterize oADSCs during in vitro expansion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service