Skip to Content
Merck
All Photos(1)

Key Documents

AB9084

Sigma-Aldrich

Anti-Polycystin-L Antibody

serum, Chemicon®

Synonym(s):

Anti-PCL, Anti-PKD2L, Anti-PKDL, Anti-TRPP3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Pkd2(18764)

Specificity

Polycystin-L

Immunogen

Synthetic peptide from the N-terminal of human Polycystin-L1.

Application

Research Category
Metabolism
Research Sub Category
Renal Physiology
This Anti-Polycystin-L Antibody is validated for use in WB, IC, IH for the detection of Polycystin-L.
Western blot: 1:1,000 using ECL. The antibody reacts with the 90 kDa Polycystin1-L1 protein. Suggested blocking buffer is 10% normal goat serum (or same host as your secondary antibody), 1% BSA in 0.1M PBS with 0.05% Tween 20. Suggested dilution buffer is 1% normal goat serum (or same host as your secondary antibody), 1% BSA in 0.1M PBS with 0.05% Tween 20. Preferred gel percentage is 4-12% gradient gel.

Immunocytochemistry: 1:1,000

Immunohistochemistry: 1:1,000 overnight at 2-8°C using a fluorescently labeled secondary antibody. Suggested fixative is 4% paraformaldehyde in 0.1M PBS (one hour). Suggested permeablization method is 0.05% Triton X-100 in dilution buffer. Suggested blocking buffer is 10% normal goat serum (or same host as your secondary antibody) and 1% BSA in 0.1M PBS. Suggested dilution buffer is 1% normal goat serum (or same host as your secondary antibody) and 1% BSA in 0.1M PBS.

Optimal working dilutions must be determined by the end user.

Analysis Note

Control
POSITIVE CONTROL: adult mouse retina

NEGATIVE CONTROL: adult mouse lung

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiang Su et al.
Nature communications, 12(1), 4871-4871 (2021-08-13)
The heteromeric complex between PKD1L3, a member of the polycystic kidney disease (PKD) protein family, and PKD2L1, also known as TRPP2 or TRPP3, has been a prototype for mechanistic characterization of heterotetrametric TRP-like channels. Here we show that a truncated
Zongshi Lu et al.
Cell reports, 24(6), 1639-1652 (2018-08-09)
High salt intake is one independent risk factor for cardiac hypertrophy. Polycystic kidney disease 2-like 1 (PKD2L1, also called TRPP3) acts as a sour sensor in taste cells, and its possible role in the cardiovascular system is unknown. Here, we
Zuzana Tonelli Gombalová et al.
The Journal of comparative neurology, 528(15), 2523-2550 (2020-03-27)
Cerebrospinal fluid contacting neurons (CSF-cNs) represent a specific class of neurons located in close vicinity of brain ventricles and central canal. In contrast with knowledge gained from other vertebrate species, we found that vast majority of CSF-cNs in the spinal
Liang Cao et al.
Frontiers in cellular neuroscience, 16, 992520-992520 (2022-09-27)
The neural stem cells (NSCs) in the ventricular-subventricular zone of the adult mammalian spinal cord may be of great benefit for repairing spinal cord injuries. However, the sources of NSCs remain unclear. Previously, we have confirmed that cerebrospinal fluid-contacting neurons
Fang Sun et al.
Diabetes, metabolic syndrome and obesity : targets and therapy, 13, 1349-1358 (2020-05-20)
High-sensitivity C-reactive protein (hs-CRP) has been regarded as a biomarker of low-degree inflammation in illness; however, whether CRP exerts its pathogenic effect on the cardiometabolic system remains unknown. Aside from the beneficial effects of metabolic surgery on cardiometabolic system, its impact

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service