Skip to Content
Merck
All Photos(6)

Key Documents

AB5675

Sigma-Aldrich

Anti-Metabotropic Glutamate Receptor 5 Antibody, pain

Chemicon®, from rabbit

Synonym(s):

Metabotropic glutamate receptor 5, Metabotropic Glutamate Receptor 5 Antibody, mGluR5, Anti-mGluR5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology), bovine (based on 100% sequence homology), primate (based on 100% sequence homology), horse (based on 100% sequence homology), feline (based on 100% sequence homology)

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GRM5(2915)

General description

Metabotropic glutamate receptor 5 or mGluR5 (UniProt Q3UVX5; also known as G protein-coupled receptor GRM5, G protein coupled receptor family C, group 1, member E) is encoded by the Grm5 (also known as Gprc1e, Mglur5, AI850523, 6430542K11Rik) gene in murine species (Gene ID 108071). The metabotropic glutamate receptors belong to a family of G protein-coupled receptors that have been divided into 3 groups based on their sequence homology, putative signal transduction mechanisms, and pharmacologic properties. mGluR1 and mGluR5 constitute Group I metabotropic glutamate receptors. mGluR5 is found on postsynaptic terminals of neurons and microglia. mGluR5 signals via Gq/G11 coupling to activate phospholipase C, resulting in calcium mobilization and activation of protein kinase C (PKC). In addition, mGluR5 is also reported to mediate signaling involving Src-family protein tyrosine kinase in a G protein-independent manner.

Specificity

This antibody will not detect Isoform 2.

Immunogen

KLH-conjugated linear peptide corresponding to the cytoplasmic domain of mouse Metabotropic Glutamate Receptor 5.

Application

Anti-Metabotropic Glutamate Receptor 5 Antibody, pain is an antibody against Metabotropic Glutamate Receptor 5 for use in IH & WB.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
Western Blotting Analysis: 0.1-0.5 μg/mL using ECL on rat brain microsomal preparation. Reacts with a band of ~132 kDa.
Immunohistochemistry: 0.1-0.2 μg/mL on rat brain and mouse brain sections fixed with 4% paraformaldehyde containing 15% saturated picric acid in 0.1M PB, pH 7.4.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Metabotropic Glutamate Receptor 5 in mouse hindbrain and rat colon tissue.

Quality

Evaluated by Western Blotting in microsomal preparation of rat brain tissue lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected Metabotropic Glutamate Receptor 5 in 10 µg of microsomal preparation of rat brain tissue lysate.

Target description

~150 kDa observed. Uncharacterized band(s) may appear in some lysates.

Linkage

Replaces: AB1596; 06-451

Physical form

ImmunoAffinity Purified
Purified rabbit polyclonal antibody in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Brain tissue

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pratyush S Suryavanshi et al.
Molecular pharmacology, 90(2), 96-105 (2016-05-28)
The delta family of ionotropic glutamate receptors consists of glutamate delta-1 (GluD1) and glutamate delta-2 receptors. We have previously shown that GluD1 knockout mice exhibit features of developmental delay, including impaired spine pruning and switch in the N-methyl-D-aspartate receptor subunit
Rocco G Gogliotti et al.
Human molecular genetics, 25(10), 1990-2004 (2016-03-05)
Rett syndrome (RS) is a neurodevelopmental disorder that shares many symptomatic and pathological commonalities with idiopathic autism. Alterations in protein synthesis-dependent synaptic plasticity (PSDSP) are a hallmark of a number of syndromic forms of autism; in the present work, we
Kalina Andreeva et al.
Ophthalmology and eye diseases, 6, 43-54 (2014-09-12)
Ischemia/reperfusion (IR) injury has been associated with several retinal pathologies, and a few genes/gene products have been linked to IR injury. However, the big picture of temporal changes, regarding the affected gene networks, pathways, and processes remains to be determined.
Laura T Haas et al.
The Journal of biological chemistry, 289(41), 28460-28477 (2014-08-26)
Soluble Amyloid-β oligomers (Aβo) can trigger Alzheimer disease (AD) pathophysiology by binding to cell surface cellular prion protein (PrP(C)). PrP(C) interacts physically with metabotropic glutamate receptor 5 (mGluR5), and this interaction controls the transmission of neurotoxic signals to intracellular substrates.
Christina Gross et al.
Cell reports, 11(5), 727-736 (2015-04-30)
The PI3K enhancer PIKE links PI3K catalytic subunits to group 1 metabotropic glutamate receptors (mGlu1/5) and activates PI3K signaling. The roles of PIKE in synaptic plasticity and the etiology of mental disorders are unknown. Here, we show that increased PIKE

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service