Skip to Content
Merck
All Photos(1)

Key Documents

AB5423

Sigma-Aldrich

Anti-Tyrosine Hydroxylase Antibody, phosphoSer31

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pSer31)

Gene Information

rat ... Th(25085)

Specificity

Tyrosine Hydroxylase, phosphoSer31. The antibody recognizes a protein of 60 kDa corresponding to TH phosphorylated at Ser31 in lysates of PC-12 cells stimulated by okadaic acid.

Immunogen

Epitope: phosphoSer31
Synthetic peptide from the phosphoSer31 of rat Tyrosine Hydroxylase. Available as catalog number AG398

Application

Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
This Anti-Tyrosine Hydroxylase Antibody, phosphoSer31 is validated for use in IC, IH, WB for the detection of Tyrosine Hydroxylase.
Western blot: 1:1,000

Immunocytochemistry: 1:1,000

Immunohistochemistry: 1:1,000

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Liquid in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA and 50% glycerol.

Storage and Stability

Maintain at -20°C in undiluted for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles. Do not store in a self defrosting freezer.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cristina Núñez et al.
Endocrinology, 148(12), 5780-5793 (2007-09-08)
Our previous studies have shown that naloxone-induced morphine withdrawal increases the hypothalamic-pituitary-adrenocortical (HPA) axis activity, which is dependent on a hyperactivity of noradrenergic pathways [nucleus tractus solitarius (NTS) A(2)] innervating the hypothalamic paraventricular nucleus (PVN). Short-term regulation of catecholamine biosynthesis
Claire B de La Serre et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 311(5), R930-R939 (2016-11-03)
Increased neuropeptide Y (NPY) gene expression in the dorsomedial hypothalamus (DMH) has been shown to cause hyperphagia, but the pathway underlying this effect remains less clear. Hypothalamic neural systems play a key role in the control of food intake, in
Cristina Núñez et al.
Endocrinology, 150(7), 3118-3127 (2009-01-31)
Chronic opiate exposure induces neurochemical adaptations in the noradrenergic system. Enhanced responsiveness of the hypothalamo-pituitary-adrenal axis after morphine withdrawal has been associated with hyperactivity of ascending noradrenergic input from the nucleus of the solitary tract (NTS-A(2)) cell group to the
Rubén García-Cabrerizo et al.
Addiction biology, 24(2), 239-250 (2017-12-29)
While prior studies have established various interacting mechanisms and neural consequences (i.e. monoaminergic nerve terminal damage) that might contribute to the adverse effects caused by methamphetamine administration, the precise mechanisms that mediate relapse during withdrawal remain unknown. This study evaluated
D García-Pérez et al.
British journal of pharmacology, 168(8), 1889-1901 (2012-12-12)
Alterations in transcription factors that regulate the development and maintenance of dopamine (DA) neurons (such as Nurr1 and Pitx3) play an important role in the pathogenesis of addiction diseases. We have examined the effects of acute and chronic morphine and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service