Skip to Content
Merck
All Photos(1)

Key Documents

AB5066P

Sigma-Aldrich

Anti-Norepinephrine Transporter Antibody

Chemicon®, from rabbit

Synonym(s):

NET

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

Specificity

Specific for Norepinephrine Transporter (NET). Norepinephrine Transporter is a 617 amino acid long integral membrane protein belonging to the Sodium:Neurotransmitter Symporter family (Bruss et al. 1995; Melikian et al. 1994; Pacholczyk et al. 1991) The immunogen peptide shows no significant homology with other other known proteins.

Immunogen

A 22 amino acid peptide sequence mapping to the 1st extracellular domain of rat NET.

Application

This Anti-Norepinephrine Transporter Antibody is validated for use in ELISA, WB for the detection of Norepinephrine Transporter.
Western blot: 1-10 μg/mL (Chemiluminescence technique): NET can exist is at least two forms, a highly glycosylated ~80kDa form is the most common in brain, and a lessly glycosylated 54kDa form is also seen in some regions and tissues.



ELISA: 1:10,000-1:100,000 (50-100 ng immunogen peptide/well) Optimal working dilutions must be determined by the end user.

Target description

80 & 54 kDa

Physical form

Affinity Purified immunoglobulin. Liquid in PBS with 0.1% BSA .

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christophe Wersinger et al.
The European journal of neuroscience, 24(11), 3141-3152 (2006-12-13)
Alpha-synuclein (alpha-Syn) has been studied in the context of Parkinson's disease, but its normative role remains elusive. We have shown that alpha-Syn regulates the homeostasis of dopaminergic and serotonergic synapses, through trafficking of the dopamine and serotonin transporter, respectively. In
Anastasia Drobysheva et al.
American journal of physiology. Heart and circulatory physiology, 305(12), H1817-H1829 (2013-10-15)
Central mechanisms involving mineralocorticoid receptor (MR) activation contribute to an increase in sympathetic tone after myocardial infarction (MI). We hypothesized that this central mechanism also contributes to cardiac sympathetic axonal sprouting and that central MR blockade reduces cardiac sympathetic hyperinnervation
Michael Y Aksenov et al.
Neurotoxicology, 29(6), 971-977 (2008-07-09)
Drug abuse is a risk factor for neurological complications in HIV infection. Cocaine has been shown to exacerbate HIV-associated brain pathology and enhance neurotoxicity of HIV-1 Tat and gp120 proteins. In this study, we found that the selective inhibitor of
X Zhang et al.
Physiological research, 64(1), 103-110 (2014-09-07)
The colorectum (late distal colon) is innervated by the sympathetic nervous system, and many colorectal diseases are related to disorders of the sympathetic nervous system. The sympathetic regulation of colorectal ion transport is rarely reported. The present study aims to
The mouse olfactory peduncle. 2.The anterior limb of the anterior commissure.
Brunjes, PC
Frontiers in Neuroanatomy null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service