Skip to Content
Merck
All Photos(4)

Key Documents

AB4336

Sigma-Aldrich

Anti-SCA-1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Lymphocyte antigen 6A-2/6E-1, Ly-6A.2/Ly-6E.1, T-cell-activating protein, Stem cell antigen 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Ly6A(110454)

General description

Stem cell antigen 1 (SCA-1) is a membrane protein belonging to the Ly-6 family of proteins and contains a single UPAR/Ly6 domain. SCA-1 is involved in the activation of T-cells and is thought to have a critical role in the differentiation, proliferation, and survival of a variety of stem cells. Recent studies have indicated SCA-1 to be an ideal marker for the isolation and purification of stem cells for research and further characterization. It is expressed by both progenitor and stem cells from several tissue types including; cardiac, hematopoietic, mammary, muscle, skin and testis.

Specificity

No Homology to human was found.
This antibody recognizes SCA-1 at the UPAR/Ly6 domain.

Immunogen

Epitope: UPAR/Ly6 domain
KLH-conjugated linear peptide corresponding to mouse SCA-1 at the UPAR/Ly6 domain.

Application

Anti-SCA-1 Antibody is an antibody against SCA-1 for use in WB & IC.
Immunocytochemistry Analysis: A 1:500 dilution from a previous lot detected SCA-1 in rat mesenchymal stem cells.
Research Category
Stem Cell Research
Research Sub Category
Hematopoietic Stem Cells

Quality

Evaluated by Western Blot in mouse lung tissue lysate.

Western Blot Analysis: 0.1 µg/mL of this antibody detected SCA-1 in 10 µg of mouse lung tissue lysate.

Target description

~14 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse lung tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Santosh K Yadav et al.
Cell death & disease, 11(3), 186-186 (2020-03-15)
Providing a conducive microenvironment is critical to increase survival of transplanted stem cells in regenerative therapy. Hyperglycemia promotes stem cell death impairing cardiac regeneration in the diabetic heart. Understanding the molecular mechanisms of high glucose-induced stem cell death is important
He Wang et al.
Bioactive materials, 16, 433-450 (2022-04-14)
Small-diameter vascular grafts fabricated from synthetic biodegradable polymers exhibit beneficial mechanical properties but often face poor regenerative potential. Different tissue engineering approaches have been employed to improve tissue regeneration in vascular grafts, but there remains a requirement for a new
Mehmet H Kural et al.
Cell transplantation, 27(9), 1375-1389 (2018-08-11)
Invasive coronary interventions can fail due to intimal hyperplasia and restenosis. Endothelial cell (EC) seeding to the vessel lumen, accelerating re-endothelialization, or local release of mTOR pathway inhibitors have helped reduce intimal hyperplasia after vessel injury. While animal models are
Gabriel Haas et al.
Journal of biomedical materials research. Part A, 109(11), 2280-2293 (2021-05-08)
Skeletal muscle is inept in regenerating after traumatic injuries such as volumetric muscle loss (VML) due to significant loss of various cellular and acellular components. Currently, there are no approved therapies for the treatment of muscle tissue following trauma. In
Christina Doherty et al.
International journal of molecular sciences, 25(3) (2024-02-10)
Peripheral nerve injury denervates muscle, resulting in muscle paralysis and atrophy. This is reversible if timely muscle reinnervation occurs. With delayed reinnervation, the muscle's reparative ability declines, and muscle-resident fibro-adipogenic progenitor cells (FAPs) proliferate and differentiate, inducing fibro-fatty muscle degradation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service