Skip to Content
Merck
All Photos(1)

Key Documents

AB3316P

Sigma-Aldrich

Anti-Monocarboxylate Transporter 4 Antibody

Chemicon®, from rabbit

Synonym(s):

MCT4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SLC16A3(9123)

Specificity

Recognizes human Monocarboxylate Transporter 4 (MCT4). The immunogen shows no significant sequence homology with other MCT. MCT4 is prominently expressed in skeletal muscle and other cells with high glycolytic rate such as tumors cells and white blood cells suggesting an important role in lactic acid efflux.

Note: According to new nomenclature, the old MCT3 has now been reclassified as MCT4. Consult Halestrap and Price (1999) Biochem. J. 343:281-299.

Immunogen

A 19 amino acid peptide sequence within the cytoplasmic domain #4 of human MCT4 (Wilson et al., 1998; Price et al., 1998).

Application

Anti-Monocarboxylate Transporter 4 Antibody detects level of Monocarboxylate Transporter 4 & has been published & validated for use in ELISA & WB.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
Western blot: 1-5 μg/mL using ECL.
Immunohistochemistry: Not tested. Recommended antibody testing dilution: 5-10 μg/mL on paraformaldehyde fixed tissue.
ELISA: 1:10,000-1:100,000 using 50-100 ng control peptide (Catalog number AG219) per well.
Optimal working dilutions must be determined by the end user.

Linkage

Replaces: AB3548P

Physical form

Affinity purified immunoglobulin. Liquid in PBS containing 0.1% BSA.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Céline Pinheiro et al.
Virchows Archiv : an international journal of pathology, 452(2), 139-146 (2008-01-12)
Tumour cells are known to be highly glycolytic, thus producing high amounts of lactic acid. Monocarboxylate transporters (MCTs), by promoting the efflux of the accumulating acids, constitute one of the most important mechanisms in the maintenance of tumour intracellular pH.
Stefan De Smet et al.
Frontiers in physiology, 8, 347-347 (2017-06-18)
In the setting of "living high," it is unclear whether high-intensity interval training (HIIT) should be performed "low" or "high" to stimulate muscular and performance adaptations. Therefore, 10 physically active males participated in a 5-week "live high-train low or high"
Importance of pH regulation and lactate/H+ transport capacity for work production during supramaximal exercise in humans.
Messonnier, L; Kristensen, M; Juel, C; Denis, C
Journal of Applied Physiology (1985)
Yi-Chen Sun et al.
Investigative ophthalmology & visual science, 58(9), 3716-3722 (2017-07-25)
To determine the presence of monocarboxylate transporter (MCT) in human and rabbit corneal epithelium and its role in transcellular fluorescein transportation in the cornea. The presence of MCTs in human and rabbit corneal epithelium was determined by RT-PCR and immunohistochemistry.
Cian McGinley et al.
Experimental physiology, 101(12), 1565-1580 (2016-10-01)
What is the central question of this study? Following a training intervention, how is the interpretation of adaptations in skeletal muscle H+ transporters influenced by biopsy timing in the context of individual protein and mRNA kinetics after the final exercise

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service