Skip to Content
Merck
All Photos(1)

Key Documents

AB3230

Sigma-Aldrich

Anti-Low Density Lipoprotein Antibody, copper oxidized

serum, Chemicon®

Synonym(s):

LDL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LDLR(3949)

Specificity

Reacts strongly to fully oxidatized modifications of LDL including Cu2+-oxidized LDL, MDA-LDL, HOCL-LDL but not to other oxidized proteins like MDA-HSA, HOCL-HSA, MDA-HDL, HOCL-HDL (below detection limit). The reaction of native LDL was weak, but clearly detectable (approx. 20%). Minimally oxidized LDL gave a strong binding signal (>80%).

Immunogen

Epitope: copper oxidized
Fully Cu2+-oxidized LDL.

Application

Anti-Low Density Lipoprotein Antibody, copper oxidized is an antibody against Low Density Lipoprotein for use in ELISA, WB, IH.
Research Category
Metabolism
Research Sub Category
Lipid Metabolism & Weight Regulation
Western Blot

Immunohistochemistry: 1:50-1:500

ELISA: 1:500-1:5,000

Optimal working dilutions must be determined by the end user.

Physical form

Rabbit serum. Lyophilized. Contains no preservative. Reconstitute with 100 μL of sterile distilled water.

Storage and Stability

Maintain lyophilized material at 2-8°C for up to 12 months. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deepa Sankaran et al.
Nephron. Experimental nephrology, 106(4), e122-e128 (2007-07-12)
Dietary soy protein and flax oil retard kidney disease progression when initiated in the early stages of disease in several experimental models, including the Han:SPRD-cy rat. However, individuals with kidney disease often do not become aware of their condition until
Malcolm R Ogborn et al.
Lipids, 43(9), 783-791 (2008-07-17)
Conjugated linoleic acid (CLA) is anti-proliferative and anti-inflammatory in the Han:SPRD-cy rat model of kidney disease. We used different doses of CLA and examined effects on renal histological benefit, the renal PPARgamma system and hepatic and renal levels of CLA
Dietary soya protein during pregnancy and lactation in rats with hereditary kidney disease attenuates disease progression in offspring.
Cahill, LE; Peng, CY; Bankovic-Calic, N; Sankaran, D; Ogborn, MR; Aukema, HM
The British Journal of Nutrition null
Tadashi Uramatsu et al.
Biological & pharmaceutical bulletin, 36(8), 1271-1277 (2013-08-02)
Stroke-prone spontaneously hypertensive (SHRsp) rats develop severe hypertension resulting in renal injury. We investigated apoptosis inhibitor of macrophages (AIM) expression in nephrosclerotic rats and the involvement of AIM in olmesartan (OLM)- and azelnidipine (AZN)-induced decreases in the number of macrophages
Deficiency in the metabolite receptor SUCNR1 (GPR91) leads to outer retinal lesions.
Favret, S; Binet, F; Lapalme, E; Leboeuf, D; Carbadillo, J; Rubic, T; Picard, E; Mawambo et al.
Aging null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service