Skip to Content
Merck
All Photos(1)

Key Documents

AABP02500

Millipore

MCE Membrane Filter, 0.8 μm Pore Size

MF-Millipore, filter diam. 25 mm, hydrophilic, black

Synonym(s):

MF-Millipore Membrane Filter, 0.8 µm pore size

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24
Pricing and availability is not currently available.

material

black filter
mixed cellulose esters (MCE) membrane
plain filter

Quality Level

description

25 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, black, 100 discs

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

MF-Millipore
Millipore

parameter

16 L/min-cm2 air flow rate
190 mL/min-cm2 water flow rate
75 °C max. temp.

filter diam.

25 mm

thickness

150 μm

gravimetric extractables

4%

color

black

matrix

MF-Millipore

pore size

0.8 μm pore size
82 % porosity

bubble point

≥1.0 bar, air with water at 23 °C

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

MF-Millipore membrane filters comprise biologically inert mixtures of cellulose acetate and cellulose nitrate. They are widely used membranes in analytical and research applications.

Application

MF-Millipore Membrane Filter, 0.8 μm pore size has been used:
  • to place retina samples before immunofluorescence staining[1]
  • in microelectrode array stimulation and recording[2]
  • in patch-clamp recording studies[3]

Features and Benefits

  • Applicable for biological and environmental monitoring.
  • Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods.

Legal Information

MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yijie Lu et al.
PloS one, 15(7), e0236176-e0236176 (2020-07-23)
Optimizing stimulation protocol is essential for clinical application of retinal prosthesis. Elongating stimulation pulse width (~25ms /phase) has been proposed as an effective method to improve spatial resolution of epi-retinal implants. However, it is unknown whether longer stimulus pulse width
Giuseppe Federighi et al.
The Journal of comparative neurology, 528(4), 624-636 (2019-10-01)
A crucial role in ejaculation is thought to be played by a population of lumbar spino-thalamic neurons (LSt), which express galanin and other neuropeptides. In rats, these neurons are activated with ejaculation and their lesion selectively abolishes ejaculation but not
Le Sun et al.
PloS one, 6(5), e19477-e19477 (2011-05-17)
The ON-OFF direction selective ganglion cells (DSGCs) in the mammalian retina code image motion by responding much more strongly to movement in one direction. They do so by receiving inhibitory inputs selectively from a particular sector of processes of the
Wen-Long Sheng et al.
PloS one, 10(2), e0117967-e0117967 (2015-02-26)
Intrinsically photosensitive retinal ganglion cells (ipRGCs) are involved in circadian and other non-image forming visual responses. An open question is whether the activity of these neurons may also be under the regulation mediated by the neurohormone melatonin. In the present

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service