Skip to Content
Merck
All Photos(1)

Key Documents

8.41863

Sigma-Aldrich

N,N-Dimethyloctylamine

for synthesis

Synonym(s):

N,N-Dimethyloctylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H23N
CAS Number:
Molecular Weight:
157.30
MDL number:
UNSPSC Code:
12352116
EC Index Number:
230-939-3
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

1.066 hPa ( 25 °C)

Quality Level

Assay

≥93.5% (GC)

form

liquid

potency

162 mg/kg LD50, oral (Rat)

refractive index

n/D 1.4243

bp

195 °C/1013 hPa

mp

-57 °C

transition temp

flash point 65 °C

density

0.765 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(CCCCCCCC)(C)C

InChI

1S/C10H23N/c1-4-5-6-7-8-9-10-11(2)3/h4-10H2,1-3H3

InChI key

UQKAOOAFEFCDGT-UHFFFAOYSA-N

Application

<ul>
<li><strong>Antimicrobial and Mutagenic Activity Research:</strong> N,N-Dimethyloctylamine is used as a versatile organic synthesis reagent used in the synthesis of quaternary diammonium salts derivatives of 1,4:3,6-dianhydro-<span class="html-small-caps">l</span>-iditol (Sikora et al., 2022).</li>
<li><strong>Antibacterial Membrane Synthesis:</strong> In the synthesis and characterization of cellulose acetate reverse osmosis membranes, which demonstrated excellent antibacterial properties. N,N-Dimethyloctylamine could be used in modifying these materials to improve their performance and durability, particularly in water purification systems (Li et al., 2019).</li>
<li><strong>Chromatographic Performance Improvement:</strong> Calabuig-Hern and aacute;ndez et al. assessed the performance of amines, including possibly N,N-Dimethyloctylamine, as silanol suppressors in reversed-phase liquid chromatography. This application underscores the compound′s role in enhancing chromatographic techniques, which are pivotal in analytical and preparative chemistry (Calabuig-Hern and aacute;ndez et al., 2016).</li>
</ul>

Analysis Note

Assay (GC, area%): ≥ 93.5 % (a/a)
Density (d 20 °C/ 4 °C): 0.764 - 0.769
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 1B - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

156.2 °F - closed cup

Flash Point(C)

69 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service