8.41808
1,2,6-Hexanetriol
for synthesis
Synonym(s):
1,2,6-Hexanetriol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H14O3
CAS Number:
Molecular Weight:
134.17
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-424-6
NACRES:
NA.22
Recommended Products
vapor pressure
0.01 hPa ( 20 °C)
Quality Level
Assay
≥96.0% (GC)
form
liquid
potency
15500 mg/kg LD50, oral (Rat)
pH
4.5-7.5 (25 °C, 300 g/L in H2O)
bp
178 °C/6.5 hPa
mp
-30 °C
transition temp
flash point 195 °C
density
1.11 g/cm3 at 20 °C
storage temp.
2-30°C
InChI
1S/C6H14O3/c7-4-2-1-3-6(9)5-8/h6-9H,1-5H2
InChI key
ZWVMLYRJXORSEP-UHFFFAOYSA-N
Application
1,2,6-Hexanetriol can be used as a reactant to synthesize:
- Unsaturated alcohols via rhenium-catalyzed deoxydehydration.
- Branched polyesters via melt polycondensation with dicarboxylic acids in the presence of tertrabutyl titanate as a catalyst.
- Shape memory polymer foams with enhanced degradation resistance.
- α-Hydroxyketones via catalytic selective oxidation of 1,2-diols.
- Primary tosylates via dibutyltin oxide-catalyzed tosylation with tosyl chloride.
Analysis Note
Assay (GC, area%): ≥ 96.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.100 - 1.107
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.
Density (d 20 °C/ 4 °C): 1.100 - 1.107
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
383.0 °F
Flash Point(C)
195 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service