Skip to Content
Merck
All Photos(1)

Key Documents

8.14674

Sigma-Aldrich

2-Mercaptobenzothiazole

for synthesis

Synonym(s):

2-Mercaptobenzothiazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5NS2
CAS Number:
Molecular Weight:
167.25
MDL number:
UNSPSC Code:
12352105
EC Index Number:
205-736-8
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

<0.000003 hPa ( 25 °C)

Quality Level

Assay

≥97.0% (acidimetric)

form

solid

autoignition temp.

465 °C

potency

1680 mg/kg LD50, oral (Rat)
>7940 mg/kg LD50, skin (Rabbit)

expl. lim.

15 % (v/v)

pH

7 (25 °C, 0.12 g/L in H2O)

bp

>260 °C (decomposes)

mp

180-183 °C

transition temp

flash point 243 °C

solubility

0.12 g/L

density

1.42 g/cm3 at 20 °C

bulk density

390 kg/m3

storage temp.

2-30°C

SMILES string

[s]1c2c([nH][c]1=S)cccc2

InChI

1S/C7H5NS2/c9-7-8-5-3-1-2-4-6(5)10-7/h1-4H,(H,8,9)

InChI key

YXIWHUQXZSMYRE-UHFFFAOYSA-N

Application

<ul>
<li><strong>DFT studies of 2-mercaptobenzothiazole and 2-mercaptobenzimidazole as corrosion inhibitors for copper:</strong> This work explores the adsorption of MBT and MBI on Cu(111) using DFT studies, highlighting its applications in corrosion inhibition ().</li> </ul>

Analysis Note

Assay (acidimetric): ≥ 97.0 %
Melting range (lower value): ≥ 176 °C
Melting range (upper value): ≤ 181 °C
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

392.0 °F - closed cup

Flash Point(C)

200 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service