Skip to Content
Merck
All Photos(1)

Key Documents

70953

Sigma-Aldrich

Rosetta Competent Cells - Novagen

Escherichia coli, rod shaped

Synonym(s):

Competent cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106202
NACRES:
NA.81

Product Name

Rosetta Competent Cells - Novagen, Rosetta host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli.

biological source

Escherichia coli

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze

growth mode

adherent or suspension

morphology

rod shaped

technique(s)

microbiological culture: suitable

cell transformation

transformation efficiency: >2×106 cfu/μg

shipped in

dry ice

storage temp.

−70°C

General description

Genotype: F-ompT hsdSB(rB- mB-) gal dcm pRARE (CamR)

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges us to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.
Rosetta host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli. These strains supply tRNAs for AGG, AGA, AUA, CUA, CCC, GGA codons on a compatible chloramphenicol-resistant plasmid. Thus the Rosetta strains provide for “universal” translation which is otherwise limited by the codon usage of E. coli. The tRNA genes are driven by their native promoters.
Rosetta host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli.

Components

0.4 ml1 mlComponent

•2 × 0.2 ml5 × 0.2 mlRosetta Competent Cells

•2 × 2 ml4 × 2 mlSOC Medium

•10 µl10 µlTest Plasmid

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yingying Yang et al.
PLoS genetics, 19(8), e1010733-e1010733 (2023-08-21)
The mitochondrial C-to-U RNA editing factor PPR56 of the moss Physcomitrium patens is an RNA-binding pentatricopeptide repeat protein equipped with a terminal DYW-type cytidine deaminase domain. Transferred into Escherichia coli, PPR56 works faithfully on its two native RNA editing targets
Erdem Sendinc et al.
Molecular cell, 75(3), 620-630 (2019-07-08)
mRNA modifications play important roles in regulating gene expression. One of the most abundant mRNA modifications is N6,2-O-dimethyladenosine (m6Am). Here, we demonstrate that m6Am is an evolutionarily conserved mRNA modification mediated by the Phosphorylated CTD Interacting Factor 1 (PCIF1), which
Bekir Altas et al.
Methods in molecular biology (Clifton, N.J.), 1538, 69-82 (2016-12-13)
The chemical synapse displays specialized intercellular adhesion between pre- and potsynaptic plasma membranes mediated by synaptic cell adhesion proteins. In this asymmetric cell adhesion, pre- and postsynapses have their own unique functions; the presynaptic terminal releases neurotransmitter, which diffuses through
Zsofia Botyanszki et al.
Biotechnology and bioengineering, 112(10), 2016-2024 (2015-05-08)
Biocatalytic transformations generally rely on purified enzymes or whole cells to perform complex transformations that are used on industrial scale for chemical, drug, and biofuel synthesis, pesticide decontamination, and water purification. However, both of these systems have inherent disadvantages related
Benjamin J Umlauf et al.
BioTechniques, 58(2), 81-84 (2015-02-06)
Amplification bias is a major hurdle in phage display protocols because it imparts additional, unintended selection pressure beyond binding to the desired target. One potential source of amplification bias is the inherent lack of codon optimization that occurs within phage

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service