Skip to Content
Merck
All Photos(1)

Key Documents

70663

Millipore

Proteinase K, Lyophilized

Highly active serine protease that exhibits broad cleavage specificity on native and denatured proteins and is widely used in the purification of DNA and RNA.

Synonym(s):

Proteinase K from Tritirachium album, Endopeptidase K

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.85

Quality Level

form

solid

mol wt

28.93 kDa

manufacturer/tradename

Novagen®

storage condition

OK to freeze

technique(s)

DNA extraction: suitable

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Proteinase K is a highly active 28,904-Daserine protease isolated from the fungusTritirachium album. The enzyme exhibitsbroad cleavage specificity on native anddenatured proteins and is widely used in thepurification of DNA and RNA. Its activityis increased in the presence of denaturantssuch as SDS (1%) and elevated temperature(50-60°C). The recommended workingconcentration is 50-100 µg/ml for proteinremoval and enzyme inactivation, and up to2 mg/ml for tissue treatment. The Proteinase K,Lyophilized powder can be prepared as a20 mg/ml stock solution in water and storedin aliquots at -20°C. The enzyme is alsoavailable as a ready-to-use concentrated stocksolution (600 mAU/ml) that is convenientfor routine use in most applications. 1 mgof Proteinase K is the equivalent of 30 mAU(AU = Anson unit). The Novagen ProteinaseK products are free of detectable DNase andRNase

Application

Useful for the proteolytic inactivation of nucleases during the isolation of DNA and RNA.
Removes endotoxins that bind to cationic proteins such as lysozyme and ribonuclease A.
Reported useful for the isolation of hepatic, yeast, and mung bean mitochondria
Determination of enzyme localization on membranes
Treatment of paraffin embedded tissue sections to expose antigen binding sites for antibody labeling.
Digestion of proteins from brain tissue samples for prions in Transmissible Spongiform Encephalopathies (TSE) research.

Biochem/physiol Actions

Proteinase K is a stable and highly reactive serine protease. Evidence from crystal and molecular structure studies indicates the enzyme belongs to the subtilisin family with an active-site catalytic triad (Asp39-His69-Ser224). It is stable in a broad range of environments: pH, buffer salts, detergents (SDS), and temperature. In the presence of 0.1-0.5% SDS, proteinase K retains activity and will digest a variety of proteins and nucleases in DNA preparations without compromising the integrity of the isolated DNA.

Warning

Toxicity: Harmful (C)

Unit Definition

One AU (AU = Anson unit) is defined as the amount of enzyme that liberates 1.0 µmol (181 µg) of tyrosine from casein per minute at pH 7.5 at 37°C.

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brooke G Pantazides et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1121, 9-17 (2019-05-15)
Sulfur and nitrogen mustards are internationally banned vesicants listed as Schedule 1 chemical agents in the Chemical Weapons Convention. These compounds are highly reactive electrophiles that form stable adducts to a variety of available amino acid residues on proteins upon
A Digaitiene et al.
Journal of applied microbiology, 112(4), 732-742 (2012-02-09)
To screen five strains of lactic acid bacteria (LAB) isolated from rye sourdoughs for the potential production of antimicrobial substances. Lactobacillus sakei KTU05-06, Pediococcus acidilactici KTU05-7, Pediococcus pentosaceus KTU05-8, KTU05-9 and KTU05-10 isolated from rye sourdoughs were investigated for the
Jai Rautela et al.
Cancer immunology research, 3(11), 1207-1217 (2015-07-23)
Metastatic progression is the major cause of breast cancer-related mortality. By examining multiple syngeneic preclinical breast cancer models in mice lacking a functional type-I interferon receptor (Ifnar1(-/-) mice), we show that host-derived type-I interferon (IFN) signaling is a critical determinant
Yang Sui et al.
Nucleic acids research, 50(12), 6890-6902 (2022-06-25)
Ribonucleotides can be incorporated into DNA during replication by the replicative DNA polymerases. These aberrant DNA subunits are efficiently recognized and removed by Ribonucleotide Excision Repair, which is initiated by the heterotrimeric enzyme RNase H2. While RNase H2 is essential
Yang Deng et al.
Cell death & disease, 13(12), 1057-1057 (2022-12-21)
Acute ischemic stroke (AIS) is a global cerebrovascular disease with high disability and mortality, which has no effective therapy. Studies have demonstrated that astrocyte-derived exosomes (ADEXs) provided neuroprotection in experimental stroke models. Nevertheless, the role of exosomes derived from oxygen-glucose-deprivation/reoxygenation-stimulated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service